Gene Transcripts

Gene Symbol
LINC00521
Entrez Gene ID
256369
Gene Name
long intergenic non-protein coding RNA 521
Gene Aliases
C14orf48, c14_5713
Chromosome Location
Chr. 14: 93997270 - 94011695 on Build GRCh38
UniGene ID
Hs.143845
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NR_024182.1 2 195
NR_024183.1 2 447
NR_024184.1 2 447
GenBank mRNA AA412651.1 - 1 165
AK097741.1 - 2 442
BC021728.2 - 2 446
BC031252.1 - 2 195
BI458986.1 - 2 451
DB446670.1 - 2 75

View Full Product Details