Gene Transcripts

Gene Symbol
AUP1
Entrez Gene ID
550
Gene Name
ancient ubiquitous protein 1
Gene Aliases
Chromosome Location
Chr. 2: 74526648 - 74529897 on Build GRCh38
UniGene ID
Hs.411480
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_181575.4 NP_853553.1 11 1439
NR_126510.1 11 1501
NR_126511.1 10 1635
XM_005264392.3 XP_005264449.1 10 1384
GenBank mRNA AF100746.1 - 11 1212
AF100753.1 - 11 1239
AF100754.1 - 10 1437
AF165515.1 - 11 1219
AI580873.1 - 2 218
AK023983.1 - 11 1279
AK055566.1 - 7 2071
AK293121.1 - 11 1258
AK304886.1 - 10 1381
BC001658.1 - 11 1364 3050204
BC033646.2 - 11 1254
CR749661.1 - 10 1372

View Full Product Details