Gene Transcripts
- Gene Symbol
- MIA
- Entrez Gene ID
- 8190
- Gene Name
- melanoma inhibitory activity
- Gene Aliases
- CD-RAP
- Chromosome Location
- Chr. 19: 40775177 - 40777493 on Build GRCh38
- UniGene ID
- Hs.646364
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001202553.1 |
NP_001189482.1
|
4, 5 |
402 |
|
NM_006533.3 |
NP_006524.1
|
3, 4 |
527 |
|
GenBank mRNA |
BC005910.1 |
- |
3, 4 |
386 |
|
BM715936.1 |
- |
3, 4 |
378 |
|
BM856478.1 |
- |
3, 4 |
384 |
|
BQ681978.1 |
- |
4, 5 |
402 |
|
BT007044.1 |
- |
3, 4 |
361 |
|
CR541648.1 |
- |
3, 4 |
361 |
|
DV462488.1 |
- |
1, 2 |
138 |
|
X75450.1 |
- |
3, 4 |
400 |
|
View Full Product Details