Gene Transcripts

Gene Symbol
KRT38
Entrez Gene ID
8687
Gene Name
keratin 38
Gene Aliases
HA8, KRTHA8, hHa8
Chromosome Location
Chr. 17: 41436369 - 41441344 on Build GRCh38
UniGene ID
Hs.248188
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_006771.3 NP_006762.3 1 881
GenBank mRNA AJ786656.1 - 1 409
BC131715.1 - 1 738 40123079

View Full Product Details