Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001193464.1 | NP_001180393.1 | 2 | 161 | |
NM_015522.3 | NP_056337.1 | 2 | 161 | ||
NM_016008.3 | NP_057092.2 | 2 | 161 | ||
XM_005264364.4 | XP_005264421.1 | 2 | 138 | ||
XM_005264365.4 | XP_005264422.1 | 2 | 138 | ||
GenBank mRNA | AF151818.1 | - | 2 | 122 | |
AK074206.1 | - | 2 | 151 | ||
AK223432.1 | - | 2 | 161 | ||
AK301489.1 | - | 2 | 151 | ||
AL050006.1 | - | 1 | 14 | ||
AY083346.1 | - | 2 | 121 | ||
BC006969.1 | - | 2 | 120 | 3828551 | |
BC016324.1 | - | 2 | 155 | 4093010 | |
BC040558.1 | - | 2 | 136 | ||
BC058823.1 | - | 2 | 128 | 4093493 | |
BG716986.1 | - | 2 | 166 | ||
BX493698.1 | - | 2 | 164 |