Gene Transcripts

Gene Symbol
SELT
Entrez Gene ID
51714
Gene Name
selenoprotein T
Gene Aliases
Chromosome Location
Chr. 3: 150603279 - 150630447 on Build GRCh38
UniGene ID
Hs.369052
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_016275.3 NP_057359.2 6 2328
GenBank mRNA AK075006.1 - 6 2301
BC026350.1 - 6 2308 4796883

View Full Product Details