Gene Transcripts
- Gene Symbol
- ZCCHC10
- Entrez Gene ID
- 54819
- Gene Name
- zinc finger CCHC-type containing 10
- Gene Aliases
- Chromosome Location
- Chr. 5: 132996985 - 133026621 on Build GRCh38
- UniGene ID
- Hs.29700
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001300816.2 |
NP_001287745.1
|
3, 4 |
341 |
|
NM_001300817.2 |
NP_001287746.1
|
3, 4 |
341 |
|
NM_001308127.1 |
NP_001295056.1
|
3, 4 |
371 |
|
NM_017665.3 |
NP_060135.1
|
2, 3 |
275 |
|
NR_125337.2 |
|
2, 3 |
279 |
|
GenBank mRNA |
AF086446.1 |
- |
1, 2 |
157 |
|
AK000101.1 |
- |
2, 3 |
219 |
|
AK122970.1 |
- |
3, 4 |
286 |
|
BC005211.1 |
- |
2, 3 |
232 |
3961163 |
BC022443.1 |
- |
3, 4 |
282 |
4285562 |
BI826979.1 |
- |
2, 3 |
275 |
|
BM470321.1 |
- |
2, 3 |
226 |
|
DA905071.1 |
- |
3, 4 |
316 |
|
View Full Product Details