Gene Transcripts

Gene Symbol
PIDD1
Entrez Gene ID
55367
Gene Name
p53-induced death domain protein 1
Gene Aliases
LRDD, PIDD
Chromosome Location
Chr. 11: 799179 - 809504 on Build GRCh38
UniGene ID
Hs.592290
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_145886.3 NP_665893.2 4 870
NM_145887.3 NP_665894.2 4 870
XM_005253005.4 XP_005253062.1 5 1808
XM_005253006.4 XP_005253063.1 5 1371
XM_005253007.3 XP_005253064.1 4 888
XM_005253008.4 XP_005253065.1 5 1868
XM_011520209.2 XP_011518511.1 4 1106
XM_011520210.2 XP_011518512.1 5 1808
XM_011520211.2 XP_011518513.1 5 1808
XM_011520212.1 XP_011518514.1 4 948
XM_017017993.1 XP_016873482.1 1 302
GenBank mRNA AB208832.1 - 2 1674
AB208949.1 - 2 1638
AB209529.1 - 2 1780
AF229178.1 - 3 1072
AF274972.1 - 4 866
AF465246.1 - 1 250
AL833849.1 - 1 106
BC014904.1 - 3 808 3908102
BQ924290.1 - 1 143
BX349115.2 - 2 223

View Full Product Details