Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001037806.3 | NP_001032895.2 | 8 | 1773 | |
XM_005269050.3 | XP_005269107.1 | 8 | 2098 | ||
XM_006719525.2 | XP_006719588.1 | 8 | 2068 | ||
XM_011538610.2 | XP_011536912.1 | 9 | 1852 | ||
XM_011538611.2 | XP_011536913.1 | 9 | 1822 | ||
GenBank mRNA | AB046822.1 | - | 1 | 714 | |
BC033253.1 | - | 1 | 289 |