Gene Transcripts

Gene Symbol
ZMAT1
Entrez Gene ID
84460
Gene Name
zinc finger matrin-type 1
Gene Aliases
Chromosome Location
Chr. X: 101882288 - 101932135 on Build GRCh38
UniGene ID
Hs.496512
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001011657.3 NP_001011657.2 6 942
NM_001282400.1 NP_001269329.1 9 1498
NM_001282401.1 NP_001269330.1 8 1445
XM_005262212.4 XP_005262269.2 6 959
XM_005262213.4 XP_005262270.2 6 956
XM_005262214.3 XP_005262271.2 5 890
XM_005262216.4 XP_005262273.2 6 661
XM_006724711.3 XP_006724774.1 2 421
XM_017029903.1 XP_016885392.1 6 658
XM_017029904.1 XP_016885393.1 6 2711
GenBank mRNA AB058692.1 - 1 3575
AK094141.1 - 9 1459
AK296520.1 - 8 1410
BC140920.1 - 1 3541 9056732
BC152470.1 - 1 3575 8862607

View Full Product Details