Gene Transcripts

Gene Symbol
DPPA5
Entrez Gene ID
340168
Gene Name
developmental pluripotency associated 5
Gene Aliases
ESG1
Chromosome Location
Chr. 6: 73353062 - 73354276 on Build GRCh38
UniGene ID
Hs.125331
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001025290.2 NP_001020461.1 1, 2 174
GenBank mRNA BC137549.1 - 1, 2 174
BC137558.1 - 1, 2 174 9021558
BX092581.1 - 1, 2 129

View Full Product Details