Gene Transcripts

Gene Symbol
SP9
Entrez Gene ID
100131390
Gene Name
Sp9 transcription factor
Gene Aliases
ZNF990
Chromosome Location
Chr. 2: 174335093 - 174337540 on Build GRCh38
UniGene ID
Hs.516603
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001145250.1 NP_001138722.1 2 488

View Full Product Details