Gene Transcripts
- Gene Symbol
- PRB2
- Entrez Gene ID
- 653247
- Gene Name
- proline rich protein BstNI subfamily 2
- Gene Aliases
- IB-9, PRPPRB1, Ps, cP7
- Chromosome Location
- Chr. 12: 11391540 - 11395564 on Build GRCh38
- UniGene ID
- Hs.654486
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_006248.3 |
NP_006239.3
|
3 |
1155 |
|
GenBank mRNA |
K03208.1 |
- |
1 |
625 |
|
- Gene Symbol
- PRB1
- Entrez Gene ID
- 5542
- Gene Name
- proline rich protein BstNI subfamily 1
- Gene Aliases
- PM, PMF, PMS, PRB1L, PRB1M
- Chromosome Location
- Chr. 12: 11351823 - 11355590 on Build GRCh38
- UniGene ID
- Hs.631726
- Species
- Homo sapiens
View Full Product Details