Gene Transcripts

Gene Symbol
FAM24A
Entrez Gene ID
118670
Gene Name
family with sequence similarity 24 member A
Gene Aliases
Chromosome Location
Chr. 10: 122910595 - 122913111 on Build GRCh38
UniGene ID
Hs.369829
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001029888.1 NP_001025059.1 3 380
XM_017015638.1 XP_016871127.1 3 483
XM_017015639.1 XP_016871128.1 3 388

View Full Product Details