Gene Transcripts

Gene Symbol
ZNF792
Entrez Gene ID
126375
Gene Name
zinc finger protein 792
Gene Aliases
Chromosome Location
Chr. 19: 34956354 - 34964049 on Build GRCh38
UniGene ID
Hs.50405
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_175872.4 NP_787068.3 2, 3 546
GenBank mRNA AK304550.1 - 2, 3 461
AL517807.3 - 2, 3 544
DB257726.1 - 2, 3 461

View Full Product Details