Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001336.3 | NP_001327.2 | 4 | 737 | |
GenBank mRNA | AF009923.1 | - | 4 | 581 | |
AF073890.1 | - | 4 | 707 | ||
AF136273.1 | - | 4 | 736 | ||
AK310046.1 | - | 1 | 367 | ||
AK314931.1 | - | 4 | 743 | ||
BC042168.1 | - | 4 | 720 | 5018854 | |
BT019915.1 | - | 4 | 611 |