Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001383.3 | NP_001374.3 | 6 | 607 | |
GenBank mRNA | AF321876.1 | - | 6 | 607 | |
AK297947.1 | - | 5 | 625 | ||
AK314015.1 | - | 6 | 593 | ||
BC003099.1 | - | 6 | 569 | ||
BC096088.1 | - | 5 | 463 | 40000799 | |
BT019878.1 | - | 4 | 349 | ||
S81752.1 | - | 6 | 642 | ||
U34880.1 | - | 6 | 642 |