Gene Transcripts

Gene Symbol
RNF44
Entrez Gene ID
22838
Gene Name
ring finger protein 44
Gene Aliases
Chromosome Location
Chr. 5: 176526699 - 176543456 on Build GRCh38
UniGene ID
Hs.434888
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_014901.4 NP_055716.1 2 539
XM_005265840.2 XP_005265897.1 2 251
XM_005265841.4 XP_005265898.1 2 496
XM_005265842.4 XP_005265899.1 2 545
XM_005265843.2 XP_005265900.1 2 251
XM_005265844.3 XP_005265901.1 2 353
XM_005265845.4 XP_005265902.1 2 491
XM_006714831.2 XP_006714894.1 2 251
XM_006714832.2 XP_006714895.1 2 251
XM_011534466.2 XP_011532768.1 2 254
XM_011534467.2 XP_011532769.1 2 546
GenBank mRNA AB029023.1 - 2 424
AK302385.1 - 2 353
BC039833.1 - 2 495 5535911
BC063297.1 - 2 540 30349418

View Full Product Details