Gene Transcripts
- Gene Symbol
- ATP11B
- Entrez Gene ID
- 23200
- Gene Name
- ATPase phospholipid transporting 11B (putative)
- Gene Aliases
- ATPIF, ATPIR
- Chromosome Location
- Chr. 3: 182793503 - 182923010 on Build GRCh38
- UniGene ID
- Hs.478429
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_014616.2 |
NP_055431.1
|
18, 19 |
2303 |
|
XM_005247241.2 |
XP_005247298.1
|
18, 19 |
2302 |
|
XM_005247243.2 |
XP_005247300.1
|
17, 18 |
1974 |
|
XM_011512593.2 |
XP_011510895.1
|
18, 19 |
2302 |
|
XM_011512594.2 |
XP_011510896.1
|
18, 19 |
2302 |
|
XM_011512597.2 |
XP_011510899.1
|
16, 17 |
1869 |
|
XM_017006008.1 |
XP_016861497.1
|
16, 17 |
1866 |
|
XM_017006009.1 |
XP_016861498.1
|
18, 19 |
2072 |
|
GenBank mRNA |
AB023173.1 |
- |
5, 6 |
533 |
|
AK093727.1 |
- |
6, 7 |
775 |
|
AK296750.1 |
- |
8, 9 |
892 |
|
AK304855.1 |
- |
8, 9 |
1196 |
|
BC010630.1 |
- |
4, 5 |
431 |
|
View Full Product Details