Gene Transcripts
- Gene Symbol
- C20orf27
- Entrez Gene ID
- 54976
- Gene Name
- chromosome 20 open reading frame 27
- Gene Aliases
- Chromosome Location
- Chr. 20: 3753499 - 3768388 on Build GRCh38
- UniGene ID
- Hs.274422
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001039140.2 |
NP_001034229.1
|
4, 5 |
609 |
|
NM_001258429.1 |
NP_001245358.1
|
4, 5 |
534 |
|
NM_001258430.1 |
NP_001245359.1
|
4, 5 |
988 |
|
NR_047675.1 |
|
4, 5 |
678 |
|
GenBank mRNA |
AK000557.1 |
- |
4, 5 |
473 |
|
AK024220.1 |
- |
4, 5 |
473 |
|
AK097645.1 |
- |
4, 5 |
1116 |
|
AK125202.1 |
- |
2, 3 |
1442 |
|
AK225109.1 |
- |
4, 5 |
473 |
|
AK290955.1 |
- |
4, 5 |
678 |
|
BC012196.1 |
- |
4, 5 |
460 |
2822218 |
BC024036.1 |
- |
4, 5 |
471 |
3510659 |
BF978883.1 |
- |
4, 5 |
535 |
|
BG474804.1 |
- |
4, 5 |
528 |
|
View Full Product Details