Gene Transcripts

Gene Symbol
PPP2R2C
Entrez Gene ID
5522
Gene Name
protein phosphatase 2 regulatory subunit Bgamma
Gene Aliases
B55-GAMMA, IMYPNO, IMYPNO1, PR52, PR55G
Chromosome Location
Chr. 4: 6320578 - 6563600 on Build GRCh38
UniGene ID
Hs.479069
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001206994.1 NP_001193923.1 4, 5 401
NM_001206995.1 NP_001193924.1 4, 5 505
NM_001206996.1 NP_001193925.1 4, 5 555
NM_020416.3 NP_065149.2 3, 4 693
NM_181876.2 NP_870991.1 3, 4 347
XM_011513495.1 XP_011511797.1 3, 4 442
XM_011513496.1 XP_011511798.1 3, 4 457
GenBank mRNA AB209183.1 - 2, 3 3969
AF086924.1 - 3, 4 377
AF145027.1 - 3, 4 540
AK093115.1 - 3, 4 347
AK296474.1 - 4, 5 401
AK316003.1 - 5, 6 515
BC032954.2 - 4, 5 555 4823280
BM551976.1 - 693

View Full Product Details