Gene Transcripts

Gene Symbol
MAPK11
Entrez Gene ID
5600
Gene Name
mitogen-activated protein kinase 11
Gene Aliases
P38B, P38BETA2, PRKM11, SAPK2, SAPK2B, p38-2, p38Beta
Chromosome Location
Chr. 22: 50263713 - 50270393 on Build GRCh38
UniGene ID
Hs.57732
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_002751.6 NP_002742.3 9 833
NR_110887.1 9 833
XM_011530706.1 XP_011529008.1 7 557
GenBank mRNA AF001008.1 - 9 732
AF001174.1 - 9 738
AF031135.1 - 9 747
AK291845.1 - 9 784
AK299745.1 - 6 1301
AK308969.1 - 9 749
AK309046.1 - 7 1491
BC027933.1 - 9 790 5199628
CR456514.1 - 9 734
U53442.1 - 9 775
U92268.1 - 9 736
Y14440.1 - 9 738

View Full Product Details