Gene Transcripts

Gene Symbol
CWC22
Entrez Gene ID
57703
Gene Name
CWC22 homolog, spliceosome-associated protein
Gene Aliases
EIF4GL, NCM, fSAPb
Chromosome Location
Chr. 2: 179944876 - 180007359 on Build GRCh38
UniGene ID
Hs.311363
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020943.2 NP_065994.1 7 959
XM_005246726.2 XP_005246783.1 7 1062
GenBank mRNA AB046824.1 - 6 778
AK025635.1 - 7 816
BC016651.1 - 6 783
BC031216.1 - 7 959
BC053573.1 - 4 481
BC057826.1 - 7 817
BC093952.1 - 6 681 7939797
BC093954.1 - 6 681 7939799
BC143433.1 - 6 680

View Full Product Details