Gene Transcripts

Gene Symbol
NTN4
Entrez Gene ID
59277
Gene Name
netrin 4
Gene Aliases
PRO3091
Chromosome Location
Chr. 12: 95657807 - 95790758 on Build GRCh38
UniGene ID
Hs.201034
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_021229.3 NP_067052.2 10 2249
GenBank mRNA AF119916.3 - 11 1786
AF278532.1 - 10 2251
AF297711.1 - 10 2101
AK024691.1 - 9 1325
AK026833.1 - 5 595
AK226070.1 - 10 2106
AK290591.1 - 10 1786
AK315796.1 - 10 2108
AL137540.1 - 8 1117
AL833767.1 - 9 1653
AY330211.1 - 9 1731
BC013591.1 - 9 1324
BC136798.1 - 10 2224 9020788
BC136799.1 - 10 2224 9020789

View Full Product Details