Gene Transcripts

Gene Symbol
C8B
Entrez Gene ID
732
Gene Name
complement component 8, beta polypeptide
Gene Aliases
C82
Chromosome Location
Chr. 1: 56929210 - 56974383 on Build GRCh38
UniGene ID
Hs.391835
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_000066.3 NP_000057.2 8 1401
NM_001278543.1 NP_001265472.1 9 1623
NM_001278544.1 NP_001265473.1 9 1530
XM_017002235.1 XP_016857724.1 9 1469
GenBank mRNA AK292740.1 - 8 1256
AK298425.1 - 9 1447
AK298444.1 - 9 1623
AK313382.1 - 8 1273
AK316209.1 - 9 1620
BC130575.1 - 8 1234 40146606
M16973.1 - 8 1236
X04393.1 - 7 1071

View Full Product Details