Gene Transcripts

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
Chromosome Location
Chr. 2: 97712030 - 97744327 on Build GRCh38
UniGene ID
Hs.234569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079.3 NP_001070.2 11, 12 1684
NM_207519.1 NP_997402.1 3, 4 702
XM_017004867.1 XP_016860356.1 13, 14 3085
XM_017004868.1 XP_016860357.1 13, 14 3067
XM_017004869.1 XP_016860358.1 13, 14 3085
XM_017004870.1 XP_016860359.1 13, 14 3085
GenBank mRNA AB083211.1 - 3, 4 702
AL832287.1 - 4, 5 2529
BC039039.1 - 10, 11 1338 5168073
BC053878.1 - 11, 12 1660 5757161
L05148.1 - 11, 12 1686

View Full Product Details