Gene Transcripts

Gene Symbol
ZAP70
Entrez Gene ID
7535
Gene Name
zeta chain of T cell receptor associated protein kinase 70
Gene Aliases
ADMIO2, IMD48, SRK, STCD, STD, TZK, ZAP-70
Chromosome Location
Chr. 2: 97712030 - 97744327 on Build GRCh38
UniGene ID
Hs.234569
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001079.3 NP_001070.2 14 1988
NM_207519.1 NP_997402.1 6 1006
XM_017004867.1 XP_016860356.1 16 3389
XM_017004868.1 XP_016860357.1 16 3371
GenBank mRNA AB083211.1 - 6 1006
AL832287.1 - 7 2833
BC039039.1 - 13 1642 5168073
BC053878.1 - 14 1964 5757161
L05148.1 - 14 1990

View Full Product Details