Gene Transcripts
- Gene Symbol
- IQCA1
- Entrez Gene ID
- 79781
- Gene Name
- IQ motif containing with AAA domain 1
- Gene Aliases
- 4930465P12Rik, DRC11, IQCA
- Chromosome Location
- Chr. 2: 236324147 - 236507558 on Build GRCh38
- UniGene ID
- Hs.591594
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001270584.1 |
NP_001257513.1
|
3, 4 |
724 |
|
NM_001270585.1 |
NP_001257514.1
|
3, 4 |
512 |
|
NM_024726.4 |
NP_079002.3
|
3, 4 |
724 |
|
NR_073043.1 |
|
3, 4 |
724 |
|
XM_011511864.1 |
XP_011510166.1
|
3, 4 |
510 |
|
XM_011511865.2 |
XP_011510167.1
|
3, 4 |
747 |
|
XM_011511866.1 |
XP_011510168.1
|
3, 4 |
510 |
|
XM_017004960.1 |
XP_016860449.1
|
3, 4 |
510 |
|
GenBank mRNA |
AK054711.1 |
- |
3, 4 |
511 |
|
AK294105.1 |
- |
3, 4 |
647 |
|
AK295566.1 |
- |
3, 4 |
509 |
|
BC028699.1 |
- |
3, 4 |
727 |
4839325 |
BC043504.1 |
- |
3, 4 |
638 |
5193742 |
BX117697.1 |
- |
2, 3 |
189 |
|
View Full Product Details