Gene Transcripts

Gene Symbol
VANGL1
Entrez Gene ID
81839
Gene Name
VANGL planar cell polarity protein 1
Gene Aliases
KITENIN, LPP2, STB2, STBM2
Chromosome Location
Chr. 1: 115641953 - 115698224 on Build GRCh38
UniGene ID
Hs.515130
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001172411.1 NP_001165882.1 8 1729
NM_001172412.1 NP_001165883.1 8 1679
NM_138959.2 NP_620409.1 8 1735
GenBank mRNA AB057596.1 - 8 1679
AB075805.1 - 7 1476
AF481859.1 - 8 1691
BC032773.1 - 6 1403
BC065272.1 - 8 1712 6165098

View Full Product Details