Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 72238920-72239428 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPPL1
Entrez Gene ID
3636
Gene Name
inositol polyphosphate phosphatase like 1
Gene Aliases
OPSMD, SHIP2
UniGene
Hs.523875
Gene Chromosome Location
Chr.11: 72223861-72239147 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1 NM_005169.3

Gene Symbol
PHOX2A
Entrez Gene ID
401
Gene Name
paired like homeobox 2a
Gene Aliases
ARIX, CFEOM2, FEOM2, NCAM2, PMX2A
UniGene
Hs.731115
Gene Chromosome Location
Chr.11: 72239077-72244176 on Build GRCh38
Transcripts
NM_001567.3 XM_005273978.3 XM_005273979.4 XM_011544999.2 XM_011545000.2 XM_017017673.1 NM_005169.3

SNP Detail

SNP ID Chromosome Location
rs994973273 Chr.11: 72238992-72238992 on Build GRCh38
rs950301862 Chr.11: 72238963-72238963 on Build GRCh38
rs372904530 Chr.11: 72239376-72239376 on Build GRCh38
rs930808582 Chr.11: 72239277-72239277 on Build GRCh38
rs937438585 Chr.11: 72239178-72239178 on Build GRCh38
rs571526828 Chr.11: 72239337-72239337 on Build GRCh38
rs945785030 Chr.11: 72239233-72239233 on Build GRCh38
rs1026683166 Chr.11: 72239019-72239019 on Build GRCh38
rs967313935 Chr.11: 72239404-72239404 on Build GRCh38
rs1020630899 Chr.11: 72239093-72239093 on Build GRCh38
rs770592288 Chr.11: 72239108-72239108 on Build GRCh38
rs978579896 Chr.11: 72239406-72239406 on Build GRCh38
rs998296479 Chr.11: 72239358-72239358 on Build GRCh38
rs973609888 Chr.11: 72239136-72239136 on Build GRCh38
rs1034033787 Chr.11: 72239304-72239304 on Build GRCh38
rs976659512 Chr.11: 72239069-72239069 on Build GRCh38
rs80340347 Chr.11: 72239346-72239346 on Build GRCh38
rs547019491 Chr.11: 72239339-72239339 on Build GRCh38
rs769905725 Chr.11: 72239001-72239001 on Build GRCh38
rs991996813 Chr.11: 72239179-72239179 on Build GRCh38
rs992637127 Chr.11: 72239002-72239002 on Build GRCh38
rs926009573 Chr.11: 72239166-72239166 on Build GRCh38
rs921945880 Chr.11: 72239367-72239368 on Build GRCh38
rs866844982 Chr.11: 72238993-72238993 on Build GRCh38
rs942068785 Chr.11: 72239086-72239086 on Build GRCh38
rs543154415 Chr.11: 72238979-72238979 on Build GRCh38
rs888233919 Chr.11: 72239038-72239038 on Build GRCh38
rs907862987 Chr.11: 72239061-72239061 on Build GRCh38
rs70940830 Chr.11: 72239092-72239095 on Build GRCh38
rs1014187008 Chr.11: 72239317-72239318 on Build GRCh38
rs368490749 Chr.11: 72239032-72239032 on Build GRCh38
rs967197977 Chr.11: 72239133-72239133 on Build GRCh38
rs745553282 Chr.11: 72239323-72239323 on Build GRCh38
rs1055824132 Chr.11: 72238943-72238943 on Build GRCh38
rs917234358 Chr.11: 72239208-72239208 on Build GRCh38
rs958508484 Chr.11: 72238994-72238994 on Build GRCh38
rs978632369 Chr.11: 72239150-72239150 on Build GRCh38
rs115908947 Chr.11: 72239041-72239041 on Build GRCh38
rs70940824 Chr.11: 72238968-72238968 on Build GRCh38
rs950096876 Chr.11: 72239224-72239224 on Build GRCh38
rs896044214 Chr.11: 72238961-72238961 on Build GRCh38
rs1018456787 Chr.11: 72239349-72239349 on Build GRCh38
rs951283294 Chr.11: 72239330-72239330 on Build GRCh38
rs932250524 Chr.11: 72239156-72239158 on Build GRCh38
rs888181338 Chr.11: 72239310-72239310 on Build GRCh38
rs1036581889 Chr.11: 72239240-72239240 on Build GRCh38
rs527993875 Chr.11: 72239352-72239352 on Build GRCh38
rs550093403 Chr.11: 72239252-72239252 on Build GRCh38
rs56787483 Chr.11: 72239366-72239366 on Build GRCh38
rs781538687 Chr.11: 72238955-72238956 on Build GRCh38
rs1017966966 Chr.11: 72239045-72239045 on Build GRCh38
rs144478961 Chr.11: 72239010-72239010 on Build GRCh38
rs965427802 Chr.11: 72239056-72239056 on Build GRCh38
rs1049241401 Chr.11: 72239301-72239301 on Build GRCh38
rs922166885 Chr.11: 72239147-72239147 on Build GRCh38
rs561489109 Chr.11: 72239156-72239156 on Build GRCh38
rs897954203 Chr.11: 72238983-72238983 on Build GRCh38

View Full Product Details