Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75400751-75401248 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
UniGene
Hs.546286
Gene Chromosome Location
Chr.11: 75399491-75422302 on Build GRCh38
Transcripts
NM_001005.4 NM_001256802.1 NM_001260506.1 NM_001260507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1322158 c.119G>A p.R40Q Chr.11: 75400782-75400782 on Build GRCh38
931981 c.89C>A p.A30D Chr.11: 75400752-75400752 on Build GRCh38
6400192 c.106G>C p.G36R Chr.11: 75400769-75400769 on Build GRCh38
1704399 c.127C>T p.P43S Chr.11: 75400790-75400790 on Build GRCh38
346324 c.105T>C p.S35S Chr.11: 75400768-75400768 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1018128225 Chr.11: 75400877-75400878 on Build GRCh38
rs1011588488 Chr.11: 75401046-75401046 on Build GRCh38
rs956095947 Chr.11: 75400901-75400901 on Build GRCh38
rs1028774182 Chr.11: 75401130-75401130 on Build GRCh38
rs770096413 Chr.11: 75401177-75401177 on Build GRCh38
rs17887020 Chr.11: 75400984-75400984 on Build GRCh38
rs755628828 Chr.11: 75400808-75400808 on Build GRCh38
rs985015310 Chr.11: 75400971-75400971 on Build GRCh38
rs999521852 Chr.11: 75401081-75401081 on Build GRCh38
rs11546226 Chr.11: 75400807-75400807 on Build GRCh38
rs745933807 Chr.11: 75400853-75400853 on Build GRCh38
rs528579322 Chr.11: 75400844-75400844 on Build GRCh38
rs188910119 Chr.11: 75401193-75401193 on Build GRCh38
rs1006763145 Chr.11: 75400868-75400868 on Build GRCh38
rs547968479 Chr.11: 75401145-75401145 on Build GRCh38
rs17878463 Chr.11: 75400921-75400921 on Build GRCh38
rs772877995 Chr.11: 75400850-75400850 on Build GRCh38
rs766895283 Chr.11: 75400794-75400794 on Build GRCh38
rs903311460 Chr.11: 75401065-75401065 on Build GRCh38
rs772941857 Chr.11: 75400881-75400882 on Build GRCh38
rs547324227 Chr.11: 75400898-75400898 on Build GRCh38
rs35358143 Chr.11: 75401203-75401204 on Build GRCh38
rs975864484 Chr.11: 75400942-75400942 on Build GRCh38
rs371302270 Chr.11: 75400795-75400795 on Build GRCh38
rs977679186 Chr.11: 75400885-75400885 on Build GRCh38
rs960225185 Chr.11: 75401218-75401218 on Build GRCh38
rs11546210 Chr.11: 75400818-75400818 on Build GRCh38
rs968649149 Chr.11: 75401175-75401175 on Build GRCh38
rs896837085 Chr.11: 75400834-75400834 on Build GRCh38
rs2070763 Chr.11: 75401093-75401093 on Build GRCh38
rs921675922 Chr.11: 75400943-75400943 on Build GRCh38
rs897385893 Chr.11: 75401005-75401005 on Build GRCh38
rs1030943731 Chr.11: 75401112-75401112 on Build GRCh38
rs997164889 Chr.11: 75401125-75401125 on Build GRCh38
rs543784236 Chr.11: 75401194-75401194 on Build GRCh38
rs909038714 Chr.11: 75401141-75401141 on Build GRCh38
rs1031088066 Chr.11: 75400897-75400897 on Build GRCh38
rs984531842 Chr.11: 75401140-75401140 on Build GRCh38
rs184429049 Chr.11: 75401029-75401029 on Build GRCh38
rs768300383 Chr.11: 75400987-75400987 on Build GRCh38
rs931825664 Chr.11: 75400945-75400945 on Build GRCh38
rs775768605 Chr.11: 75400865-75400865 on Build GRCh38
rs758818643 Chr.11: 75400833-75400833 on Build GRCh38
rs751440961 Chr.11: 75400777-75400777 on Build GRCh38
rs748956027 Chr.11: 75400831-75400831 on Build GRCh38
rs747365865 Chr.11: 75400878-75400881 on Build GRCh38
rs535042729 Chr.11: 75401024-75401024 on Build GRCh38
rs3206671 Chr.11: 75400815-75400815 on Build GRCh38
rs965741145 Chr.11: 75401095-75401095 on Build GRCh38
rs3133188 Chr.11: 75400993-75400993 on Build GRCh38
rs994208949 Chr.11: 75400837-75400837 on Build GRCh38
rs968047622 Chr.11: 75400846-75400846 on Build GRCh38
rs534690645 Chr.11: 75401098-75401098 on Build GRCh38
rs938829592 Chr.11: 75400970-75400970 on Build GRCh38
rs762052892 Chr.11: 75400847-75400847 on Build GRCh38
rs746604800 Chr.11: 75400864-75400864 on Build GRCh38
rs888191005 Chr.11: 75400863-75400863 on Build GRCh38
rs1043637045 Chr.11: 75401063-75401063 on Build GRCh38
rs550015312 Chr.11: 75401012-75401012 on Build GRCh38
rs778292583 Chr.11: 75400835-75400835 on Build GRCh38
rs1026762758 Chr.11: 75400862-75400862 on Build GRCh38
rs771591496 Chr.11: 75400838-75400838 on Build GRCh38
rs953119741 Chr.11: 75401132-75401132 on Build GRCh38
rs989198443 Chr.11: 75400917-75400917 on Build GRCh38
rs200779146 Chr.11: 75400836-75400836 on Build GRCh38
rs563927352 Chr.11: 75401195-75401195 on Build GRCh38
rs111591514 Chr.11: 75401023-75401023 on Build GRCh38
rs1031308973 Chr.11: 75400894-75400894 on Build GRCh38
rs972432244 Chr.11: 75401171-75401171 on Build GRCh38
rs1006648328 Chr.11: 75401025-75401025 on Build GRCh38
rs938684464 Chr.11: 75401021-75401021 on Build GRCh38
rs373592484 Chr.11: 75401162-75401162 on Build GRCh38
rs549504792 Chr.11: 75400828-75400828 on Build GRCh38
rs976687342 Chr.11: 75400893-75400893 on Build GRCh38
rs574093870 Chr.11: 75401209-75401209 on Build GRCh38
rs947464221 Chr.11: 75400923-75400923 on Build GRCh38
rs757191814 Chr.11: 75400789-75400789 on Build GRCh38
rs569313602 Chr.11: 75401017-75401017 on Build GRCh38

View Full Product Details