Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 75402023-75402562 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
RPS3
Entrez Gene ID
6188
Gene Name
ribosomal protein S3
Gene Aliases
S3
UniGene
Hs.546286
Gene Chromosome Location
Chr.11: 75399491-75422302 on Build GRCh38
Transcripts
NM_001005.4 NM_001256802.1 NM_001260506.1 NM_001260507.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4777956 c.256-3A>G p.? Chr.11: 75402349-75402349 on Build GRCh38
6998123 c.311C>A p.S104Y Chr.11: 75402407-75402407 on Build GRCh38
6989582 c.300C>T p.A100A Chr.11: 75402396-75402396 on Build GRCh38
6347334 c.309G>T p.E103D Chr.11: 75402405-75402405 on Build GRCh38
931982 c.315G>A p.L105L Chr.11: 75402411-75402411 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs897446171 Chr.11: 75402232-75402232 on Build GRCh38
rs11546234 Chr.11: 75402367-75402367 on Build GRCh38
rs752468542 Chr.11: 75402269-75402269 on Build GRCh38
rs181227845 Chr.11: 75402101-75402101 on Build GRCh38
rs17886478 Chr.11: 75402508-75402508 on Build GRCh38
rs922771417 Chr.11: 75402168-75402168 on Build GRCh38
rs955243667 Chr.11: 75402126-75402126 on Build GRCh38
rs917825066 Chr.11: 75402205-75402205 on Build GRCh38
rs969146465 Chr.11: 75402116-75402116 on Build GRCh38
rs868494605 Chr.11: 75402472-75402472 on Build GRCh38
rs772883502 Chr.11: 75402390-75402390 on Build GRCh38
rs545194393 Chr.11: 75402192-75402192 on Build GRCh38
rs756553267 Chr.11: 75402493-75402493 on Build GRCh38
rs370378078 Chr.11: 75402316-75402318 on Build GRCh38
rs1802703 Chr.11: 75402424-75402424 on Build GRCh38
rs924577622 Chr.11: 75402129-75402129 on Build GRCh38
rs934778525 Chr.11: 75402153-75402153 on Build GRCh38
rs770438930 Chr.11: 75402241-75402241 on Build GRCh38
rs1022833756 Chr.11: 75402112-75402112 on Build GRCh38
rs768762578 Chr.11: 75402344-75402344 on Build GRCh38
rs1048063869 Chr.11: 75402531-75402531 on Build GRCh38
rs748426216 Chr.11: 75402382-75402382 on Build GRCh38
rs528954822 Chr.11: 75402143-75402143 on Build GRCh38
rs895835486 Chr.11: 75402081-75402081 on Build GRCh38
rs17884709 Chr.11: 75402167-75402167 on Build GRCh38
rs538264377 Chr.11: 75402473-75402473 on Build GRCh38
rs543258854 Chr.11: 75402252-75402252 on Build GRCh38
rs866350524 Chr.11: 75402321-75402321 on Build GRCh38
rs937933788 Chr.11: 75402075-75402075 on Build GRCh38
rs34499053 Chr.11: 75402270-75402271 on Build GRCh38
rs565339976 Chr.11: 75402219-75402219 on Build GRCh38
rs17880385 Chr.11: 75402453-75402453 on Build GRCh38
rs766838537 Chr.11: 75402497-75402497 on Build GRCh38
rs563119520 Chr.11: 75402266-75402266 on Build GRCh38
rs763182317 Chr.11: 75402535-75402535 on Build GRCh38
rs764902063 Chr.11: 75402469-75402469 on Build GRCh38
rs573156640 Chr.11: 75402179-75402179 on Build GRCh38
rs765844657 Chr.11: 75402256-75402256 on Build GRCh38
rs759759970 Chr.11: 75402113-75402113 on Build GRCh38
rs756611381 Chr.11: 75402311-75402311 on Build GRCh38
rs1056337945 Chr.11: 75402201-75402201 on Build GRCh38
rs987805714 Chr.11: 75402145-75402145 on Build GRCh38
rs954223748 Chr.11: 75402535-75402535 on Build GRCh38
rs117378338 Chr.11: 75402288-75402288 on Build GRCh38
rs17881509 Chr.11: 75402231-75402231 on Build GRCh38
rs397844185 Chr.11: 75402510-75402510 on Build GRCh38
rs542416210 Chr.11: 75402244-75402244 on Build GRCh38
rs1038437947 Chr.11: 75402249-75402249 on Build GRCh38
rs975679619 Chr.11: 75402117-75402117 on Build GRCh38
rs991952905 Chr.11: 75402476-75402476 on Build GRCh38
rs780788225 Chr.11: 75402255-75402256 on Build GRCh38
rs759116015 Chr.11: 75402460-75402460 on Build GRCh38
rs141837939 Chr.11: 75402314-75402316 on Build GRCh38
rs777244880 Chr.11: 75402076-75402076 on Build GRCh38
rs779102234 Chr.11: 75402375-75402375 on Build GRCh38
rs776751854 Chr.11: 75402436-75402436 on Build GRCh38
rs762591390 Chr.11: 75402467-75402467 on Build GRCh38
rs946478788 Chr.11: 75402176-75402176 on Build GRCh38
rs11546215 Chr.11: 75402353-75402353 on Build GRCh38
rs369268647 Chr.11: 75402108-75402108 on Build GRCh38
rs895692591 Chr.11: 75402251-75402251 on Build GRCh38
rs138360173 Chr.11: 75402097-75402097 on Build GRCh38
rs1014245129 Chr.11: 75402087-75402087 on Build GRCh38
rs752343879 Chr.11: 75402470-75402470 on Build GRCh38
rs1047052359 Chr.11: 75402237-75402237 on Build GRCh38
rs1050030878 Chr.11: 75402186-75402186 on Build GRCh38
rs750777065 Chr.11: 75402492-75402492 on Build GRCh38
rs767884299 Chr.11: 75402482-75402482 on Build GRCh38
rs370272831 Chr.11: 75402330-75402330 on Build GRCh38
rs17880903 Chr.11: 75402177-75402177 on Build GRCh38
rs528039603 Chr.11: 75402412-75402412 on Build GRCh38
rs201507842 Chr.11: 75402508-75402508 on Build GRCh38
rs565558803 Chr.11: 75402384-75402384 on Build GRCh38
rs11546233 Chr.11: 75402366-75402366 on Build GRCh38
rs754408755 Chr.11: 75402500-75402500 on Build GRCh38
rs750876728 Chr.11: 75402300-75402300 on Build GRCh38
rs1026174480 Chr.11: 75402098-75402098 on Build GRCh38
rs143082680 Chr.11: 75402420-75402420 on Build GRCh38
rs749346137 Chr.11: 75402339-75402339 on Build GRCh38
rs557581308 Chr.11: 75402093-75402093 on Build GRCh38
rs959224743 Chr.11: 75402461-75402461 on Build GRCh38
rs143925312 Chr.11: 75402322-75402322 on Build GRCh38
rs543322681 Chr.11: 75402125-75402125 on Build GRCh38
rs376627478 Chr.11: 75402105-75402105 on Build GRCh38
rs757632649 Chr.11: 75402274-75402274 on Build GRCh38
rs888693378 Chr.11: 75402213-75402213 on Build GRCh38
rs772532026 Chr.11: 75402264-75402264 on Build GRCh38

View Full Product Details