Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 6026058-6026565 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNAB2
Entrez Gene ID
8514
Gene Name
potassium voltage-gated channel subfamily A regulatory beta subunit 2
Gene Aliases
AKR6A5, HKvbeta2, HKvbeta2.1, HKvbeta2.2, KCNA2B, KV-BETA-2
UniGene
Hs.440497
Gene Chromosome Location
Chr.1: 5992298-6101193 on Build GRCh38
Transcripts
NM_001199861.1 NM_003636.3 NM_172130.2 XM_005263514.2 XM_011542321.2 XM_011542322.2 XM_017002618.1 XM_017002619.1 XM_017002621.1

SNP Detail

SNP ID Chromosome Location
rs780423151 Chr.1: 6026471-6026471 on Build GRCh38
rs190848401 Chr.1: 6026442-6026442 on Build GRCh38
rs908834101 Chr.1: 6026543-6026543 on Build GRCh38
rs968508323 Chr.1: 6026240-6026240 on Build GRCh38
rs975865691 Chr.1: 6026282-6026282 on Build GRCh38
rs931649327 Chr.1: 6026273-6026273 on Build GRCh38
rs1036647159 Chr.1: 6026351-6026351 on Build GRCh38
rs1012513390 Chr.1: 6026405-6026405 on Build GRCh38
rs887466152 Chr.1: 6026279-6026279 on Build GRCh38
rs975896787 Chr.1: 6026485-6026485 on Build GRCh38
rs1036746335 Chr.1: 6026511-6026511 on Build GRCh38
rs753894626 Chr.1: 6026088-6026088 on Build GRCh38
rs75140054 Chr.1: 6026428-6026428 on Build GRCh38
rs898059778 Chr.1: 6026357-6026357 on Build GRCh38
rs902229593 Chr.1: 6026293-6026326 on Build GRCh38
rs967772322 Chr.1: 6026460-6026460 on Build GRCh38
rs574000349 Chr.1: 6026418-6026418 on Build GRCh38
rs754214681 Chr.1: 6026248-6026254 on Build GRCh38
rs915314308 Chr.1: 6026182-6026182 on Build GRCh38
rs551550349 Chr.1: 6026274-6026274 on Build GRCh38
rs991250131 Chr.1: 6026175-6026175 on Build GRCh38
rs919521205 Chr.1: 6026541-6026541 on Build GRCh38
rs976345644 Chr.1: 6026461-6026461 on Build GRCh38
rs867753305 Chr.1: 6026101-6026101 on Build GRCh38
rs978112942 Chr.1: 6026434-6026434 on Build GRCh38
rs1040260508 Chr.1: 6026271-6026271 on Build GRCh38
rs1015590903 Chr.1: 6026166-6026166 on Build GRCh38
rs1035712164 Chr.1: 6026107-6026107 on Build GRCh38
rs978165134 Chr.1: 6026245-6026245 on Build GRCh38
rs942067974 Chr.1: 6026507-6026507 on Build GRCh38
rs560647245 Chr.1: 6026462-6026462 on Build GRCh38
rs959396248 Chr.1: 6026155-6026155 on Build GRCh38
rs1031080673 Chr.1: 6026443-6026443 on Build GRCh38
rs968371730 Chr.1: 6026269-6026269 on Build GRCh38
rs567948528 Chr.1: 6026173-6026173 on Build GRCh38
rs539173172 Chr.1: 6026272-6026272 on Build GRCh38
rs968024384 Chr.1: 6026433-6026433 on Build GRCh38
rs545518037 Chr.1: 6026499-6026499 on Build GRCh38
rs909276982 Chr.1: 6026505-6026505 on Build GRCh38
rs41274540 Chr.1: 6026371-6026371 on Build GRCh38
rs909256548 Chr.1: 6026301-6026301 on Build GRCh38
rs1023263693 Chr.1: 6026262-6026262 on Build GRCh38
rs527980784 Chr.1: 6026096-6026096 on Build GRCh38
rs1003750051 Chr.1: 6026317-6026317 on Build GRCh38
rs1034789946 Chr.1: 6026192-6026192 on Build GRCh38
rs1003031124 Chr.1: 6026181-6026181 on Build GRCh38
rs555345551 Chr.1: 6026359-6026359 on Build GRCh38
rs150605623 Chr.1: 6026172-6026172 on Build GRCh38
rs114010715 Chr.1: 6026082-6026082 on Build GRCh38
rs114572138 Chr.1: 6026297-6026297 on Build GRCh38
rs184487389 Chr.1: 6026215-6026215 on Build GRCh38
rs1035258273 Chr.1: 6026361-6026361 on Build GRCh38
rs45622237 Chr.1: 6026251-6026251 on Build GRCh38
rs550174312 Chr.1: 6026250-6026250 on Build GRCh38
rs187268204 Chr.1: 6026424-6026424 on Build GRCh38
rs923737104 Chr.1: 6026246-6026246 on Build GRCh38
rs984744547 Chr.1: 6026509-6026509 on Build GRCh38
rs559510583 Chr.1: 6026530-6026530 on Build GRCh38
rs1004613108 Chr.1: 6026294-6026294 on Build GRCh38
rs762350811 Chr.1: 6026218-6026218 on Build GRCh38
rs545215001 Chr.1: 6026452-6026452 on Build GRCh38
rs888034688 Chr.1: 6026090-6026090 on Build GRCh38
rs188924286 Chr.1: 6026306-6026306 on Build GRCh38

View Full Product Details