Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 7793782-7794182 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PER3
Entrez Gene ID
8863
Gene Name
period circadian clock 3
Gene Aliases
FASPS3, GIG13
UniGene
Hs.162200
Gene Chromosome Location
Chr.1: 7784285-7845181 on Build GRCh38
Transcripts
NM_001289861.1 NM_001289862.1 NM_001289863.1 NM_001289864.1 NM_016831.2 XM_005263524.3 XM_011542385.2 XM_011542390.2 XM_017002723.1 XM_017002724.1 XM_017002725.1 XM_017002726.1 XM_017002727.1 XM_017002728.1 XM_017002729.1 XM_017002730.1 XM_017002731.1 XM_017002732.1 XM_017002733.1 XM_017002734.1 XM_017002735.1 XM_017002736.1 XM_017002737.1 XM_017002738.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5597364 c.622C>T p.P208S Chr.1: 7793989-7793989 on Build GRCh38
6744021 c.602A>G p.Y201C Chr.1: 7793969-7793969 on Build GRCh38
3985235 c.624T>C p.P208P Chr.1: 7793991-7793991 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs192331330 Chr.1: 7794111-7794111 on Build GRCh38
rs112178288 Chr.1: 7794142-7794142 on Build GRCh38
rs181523753 Chr.1: 7794145-7794145 on Build GRCh38
rs1045822042 Chr.1: 7793984-7793984 on Build GRCh38
rs942470356 Chr.1: 7793833-7793833 on Build GRCh38
rs754053184 Chr.1: 7794008-7794008 on Build GRCh38
rs751346081 Chr.1: 7793939-7793939 on Build GRCh38
rs202244670 Chr.1: 7794029-7794029 on Build GRCh38
rs201661449 Chr.1: 7793981-7793981 on Build GRCh38
rs967288808 Chr.1: 7794065-7794065 on Build GRCh38
rs752827946 Chr.1: 7793871-7793871 on Build GRCh38
rs751535447 Chr.1: 7793991-7793991 on Build GRCh38
rs763922771 Chr.1: 7793937-7793937 on Build GRCh38
rs747567192 Chr.1: 7794032-7794032 on Build GRCh38
rs747298171 Chr.1: 7793974-7793974 on Build GRCh38
rs780086666 Chr.1: 7793943-7793943 on Build GRCh38
rs778942800 Chr.1: 7794025-7794025 on Build GRCh38
rs762948790 Chr.1: 7793983-7793983 on Build GRCh38
rs966538044 Chr.1: 7793904-7793904 on Build GRCh38
rs201085150 Chr.1: 7794006-7794006 on Build GRCh38
rs768253718 Chr.1: 7793944-7793947 on Build GRCh38
rs768223607 Chr.1: 7794050-7794050 on Build GRCh38
rs762500903 Chr.1: 7794046-7794046 on Build GRCh38
rs137991591 Chr.1: 7793979-7793979 on Build GRCh38
rs1029091937 Chr.1: 7794100-7794100 on Build GRCh38
rs115768191 Chr.1: 7794105-7794105 on Build GRCh38
rs978499318 Chr.1: 7793890-7793890 on Build GRCh38
rs981112973 Chr.1: 7794112-7794112 on Build GRCh38
rs754982505 Chr.1: 7793955-7793955 on Build GRCh38
rs1011151784 Chr.1: 7793868-7793868 on Build GRCh38
rs774087439 Chr.1: 7794055-7794055 on Build GRCh38
rs995721569 Chr.1: 7793902-7793902 on Build GRCh38
rs746236516 Chr.1: 7793977-7793977 on Build GRCh38
rs771136768 Chr.1: 7794036-7794036 on Build GRCh38
rs1033838149 Chr.1: 7793859-7793859 on Build GRCh38
rs974861280 Chr.1: 7794098-7794098 on Build GRCh38
rs758538607 Chr.1: 7794026-7794026 on Build GRCh38
rs548766107 Chr.1: 7793808-7793808 on Build GRCh38
rs188245769 Chr.1: 7793930-7793930 on Build GRCh38
rs772430775 Chr.1: 7794015-7794015 on Build GRCh38
rs1045446670 Chr.1: 7793848-7793848 on Build GRCh38
rs1055218317 Chr.1: 7794009-7794009 on Build GRCh38
rs114300822 Chr.1: 7794007-7794007 on Build GRCh38
rs1003686349 Chr.1: 7793850-7793850 on Build GRCh38
rs553197066 Chr.1: 7794024-7794024 on Build GRCh38
rs930959626 Chr.1: 7794128-7794128 on Build GRCh38
rs906966205 Chr.1: 7793849-7793852 on Build GRCh38
rs1019390029 Chr.1: 7794047-7794047 on Build GRCh38
rs181072346 Chr.1: 7793940-7793940 on Build GRCh38
rs745505247 Chr.1: 7794040-7794040 on Build GRCh38
rs898176253 Chr.1: 7793893-7793893 on Build GRCh38
rs752619599 Chr.1: 7793914-7793914 on Build GRCh38
rs201029948 Chr.1: 7793966-7793966 on Build GRCh38
rs901161338 Chr.1: 7794066-7794066 on Build GRCh38
rs376607172 Chr.1: 7793948-7793948 on Build GRCh38
rs777206256 Chr.1: 7794039-7794039 on Build GRCh38
rs966699553 Chr.1: 7793883-7793883 on Build GRCh38
rs763894818 Chr.1: 7793990-7793990 on Build GRCh38
rs556720784 Chr.1: 7794140-7794141 on Build GRCh38
rs759268594 Chr.1: 7793980-7793980 on Build GRCh38
rs769606889 Chr.1: 7794041-7794041 on Build GRCh38
rs986581492 Chr.1: 7794096-7794096 on Build GRCh38
rs200114794 Chr.1: 7794042-7794042 on Build GRCh38
rs369203237 Chr.1: 7793962-7793962 on Build GRCh38
rs575815080 Chr.1: 7794109-7794109 on Build GRCh38
rs570385822 Chr.1: 7793861-7793861 on Build GRCh38
rs746050693 Chr.1: 7793887-7793887 on Build GRCh38
rs186997011 Chr.1: 7794107-7794107 on Build GRCh38
rs1004965704 Chr.1: 7793816-7793819 on Build GRCh38
rs11807683 Chr.1: 7794149-7794149 on Build GRCh38
rs773875104 Chr.1: 7793952-7793955 on Build GRCh38
rs1039344892 Chr.1: 7793837-7793837 on Build GRCh38
rs772505358 Chr.1: 7793965-7793965 on Build GRCh38
rs771350135 Chr.1: 7793975-7793975 on Build GRCh38
rs902781558 Chr.1: 7794035-7794035 on Build GRCh38
rs779361725 Chr.1: 7793867-7793867 on Build GRCh38
rs551252579 Chr.1: 7794044-7794044 on Build GRCh38
rs756941737 Chr.1: 7793995-7793995 on Build GRCh38
rs775201073 Chr.1: 7793982-7793982 on Build GRCh38
rs779030849 Chr.1: 7793961-7793961 on Build GRCh38
rs574554049 Chr.1: 7794103-7794103 on Build GRCh38

View Full Product Details