Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 108658601-108659100 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CORO1C
Entrez Gene ID
23603
Gene Name
coronin 1C
Gene Aliases
HCRNN4
UniGene
Hs.330384
Gene Chromosome Location
Chr.12: 108645109-108731550 on Build GRCh38
Transcripts
NM_001105237.2 NM_001276471.1 NM_014325.3 XM_017019121.1 XM_017019122.1 XM_017019123.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7083617 c.631T>G p.W211G Chr.12: 108658896-108658896 on Build GRCh38
3734597 c.454G>T p.D152Y Chr.12: 108658914-108658914 on Build GRCh38
6485906 c.723G>T p.L241L Chr.12: 108658804-108658804 on Build GRCh38
6220359 c.620A>C p.E207A Chr.12: 108658748-108658748 on Build GRCh38
310289 c.473G>T p.W158L Chr.12: 108658895-108658895 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778446633 Chr.12: 108658925-108658925 on Build GRCh38
rs756734816 Chr.12: 108658883-108658883 on Build GRCh38
rs1009419622 Chr.12: 108659012-108659013 on Build GRCh38
rs923307462 Chr.12: 108659064-108659064 on Build GRCh38
rs980968997 Chr.12: 108659018-108659019 on Build GRCh38
rs765399164 Chr.12: 108658694-108658694 on Build GRCh38
rs1032235694 Chr.12: 108659009-108659009 on Build GRCh38
rs778270031 Chr.12: 108658886-108658886 on Build GRCh38
rs199738350 Chr.12: 108658723-108658723 on Build GRCh38
rs534217868 Chr.12: 108658753-108658753 on Build GRCh38
rs888952087 Chr.12: 108658975-108658975 on Build GRCh38
rs760426637 Chr.12: 108658825-108658825 on Build GRCh38
rs547733644 Chr.12: 108658764-108658764 on Build GRCh38
rs771517007 Chr.12: 108658928-108658928 on Build GRCh38
rs745337401 Chr.12: 108658926-108658926 on Build GRCh38
rs998043139 Chr.12: 108658839-108658839 on Build GRCh38
rs909363121 Chr.12: 108659045-108659045 on Build GRCh38
rs190067519 Chr.12: 108659030-108659030 on Build GRCh38
rs538021318 Chr.12: 108658999-108658999 on Build GRCh38
rs763842145 Chr.12: 108658835-108658835 on Build GRCh38
rs749684362 Chr.12: 108658906-108658906 on Build GRCh38
rs1050532962 Chr.12: 108658931-108658931 on Build GRCh38
rs751589060 Chr.12: 108658853-108658853 on Build GRCh38
rs1036608247 Chr.12: 108659057-108659057 on Build GRCh38
rs545521139 Chr.12: 108659016-108659016 on Build GRCh38
rs143115944 Chr.12: 108658818-108658818 on Build GRCh38
rs148246664 Chr.12: 108658834-108658834 on Build GRCh38
rs756869306 Chr.12: 108658915-108658915 on Build GRCh38
rs940691545 Chr.12: 108658738-108658738 on Build GRCh38
rs991130337 Chr.12: 108659044-108659044 on Build GRCh38
rs756516563 Chr.12: 108659020-108659021 on Build GRCh38
rs574251737 Chr.12: 108658780-108658780 on Build GRCh38
rs767422590 Chr.12: 108658689-108658689 on Build GRCh38
rs746923531 Chr.12: 108658948-108658948 on Build GRCh38
rs903274820 Chr.12: 108658678-108658678 on Build GRCh38
rs377135365 Chr.12: 108658709-108658709 on Build GRCh38
rs937489433 Chr.12: 108658676-108658676 on Build GRCh38
rs149062887 Chr.12: 108658801-108658801 on Build GRCh38
rs763221290 Chr.12: 108658843-108658843 on Build GRCh38
rs958868487 Chr.12: 108659019-108659022 on Build GRCh38
rs950312546 Chr.12: 108659056-108659056 on Build GRCh38
rs556354780 Chr.12: 108659010-108659010 on Build GRCh38
rs753278155 Chr.12: 108658659-108658659 on Build GRCh38
rs35021885 Chr.12: 108658758-108658759 on Build GRCh38
rs755025882 Chr.12: 108658855-108658855 on Build GRCh38
rs574255972 Chr.12: 108658962-108658962 on Build GRCh38
rs755502319 Chr.12: 108658721-108658721 on Build GRCh38
rs574979541 Chr.12: 108659014-108659014 on Build GRCh38
rs777526999 Chr.12: 108658760-108658760 on Build GRCh38
rs753288003 Chr.12: 108658875-108658875 on Build GRCh38
rs768167585 Chr.12: 108658768-108658768 on Build GRCh38
rs116613120 Chr.12: 108658626-108658626 on Build GRCh38
rs768088134 Chr.12: 108658858-108658858 on Build GRCh38
rs779582898 Chr.12: 108658937-108658937 on Build GRCh38
rs771989893 Chr.12: 108658817-108658817 on Build GRCh38
rs773909841 Chr.12: 108658789-108658789 on Build GRCh38
rs758932141 Chr.12: 108658703-108658703 on Build GRCh38
rs776455083 Chr.12: 108658841-108658841 on Build GRCh38
rs984934660 Chr.12: 108659036-108659037 on Build GRCh38
rs751644971 Chr.12: 108658713-108658713 on Build GRCh38
rs766574976 Chr.12: 108658852-108658852 on Build GRCh38
rs879389167 Chr.12: 108659006-108659006 on Build GRCh38
rs750679783 Chr.12: 108658700-108658700 on Build GRCh38
rs775201477 Chr.12: 108658690-108658690 on Build GRCh38
rs756569073 Chr.12: 108658756-108658756 on Build GRCh38
rs143467241 Chr.12: 108658658-108658658 on Build GRCh38
rs916169079 Chr.12: 108659050-108659050 on Build GRCh38
rs374484534 Chr.12: 108659019-108659020 on Build GRCh38
rs951705755 Chr.12: 108659036-108659036 on Build GRCh38

View Full Product Details