Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 52581229-52581735 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM214A
Entrez Gene ID
56204
Gene Name
family with sequence similarity 214 member A
Gene Aliases
KIAA1370
UniGene
Hs.152385
Gene Chromosome Location
Chr.15: 52581321-52709817 on Build GRCh38
Transcripts
NM_001286495.1 NM_019600.3 NR_104457.1 XM_005254547.2 XM_005254548.2 XM_011521789.2 XM_011521790.1

SNP Detail

SNP ID Chromosome Location
rs995904585 Chr.15: 52581301-52581301 on Build GRCh38
rs977010466 Chr.15: 52581360-52581360 on Build GRCh38
rs780677385 Chr.15: 52581563-52581563 on Build GRCh38
rs767915636 Chr.15: 52581441-52581441 on Build GRCh38
rs937451444 Chr.15: 52581413-52581413 on Build GRCh38
rs74016849 Chr.15: 52581353-52581353 on Build GRCh38
rs550265137 Chr.15: 52581442-52581442 on Build GRCh38
rs984171664 Chr.15: 52581399-52581399 on Build GRCh38
rs1033174411 Chr.15: 52581316-52581316 on Build GRCh38
rs762323605 Chr.15: 52581429-52581429 on Build GRCh38
rs949939042 Chr.15: 52581440-52581440 on Build GRCh38
rs999909502 Chr.15: 52581312-52581312 on Build GRCh38
rs549136678 Chr.15: 52581548-52581548 on Build GRCh38
rs571678338 Chr.15: 52581459-52581459 on Build GRCh38
rs923424968 Chr.15: 52581564-52581564 on Build GRCh38
rs1054158230 Chr.15: 52581538-52581538 on Build GRCh38
rs1055530967 Chr.15: 52581417-52581417 on Build GRCh38
rs1005835074 Chr.15: 52581325-52581325 on Build GRCh38
rs114227387 Chr.15: 52581347-52581347 on Build GRCh38
rs1029161936 Chr.15: 52581315-52581315 on Build GRCh38
rs566021872 Chr.15: 52581630-52581630 on Build GRCh38
rs146593693 Chr.15: 52581696-52581696 on Build GRCh38
rs980666081 Chr.15: 52581373-52581373 on Build GRCh38
rs951454025 Chr.15: 52581398-52581398 on Build GRCh38
rs532503023 Chr.15: 52581544-52581544 on Build GRCh38
rs115634757 Chr.15: 52581412-52581412 on Build GRCh38
rs1005950773 Chr.15: 52581684-52581684 on Build GRCh38
rs936098185 Chr.15: 52581405-52581405 on Build GRCh38
rs745443756 Chr.15: 52581671-52581671 on Build GRCh38
rs927351655 Chr.15: 52581400-52581400 on Build GRCh38
rs998402103 Chr.15: 52581692-52581692 on Build GRCh38
rs953191678 Chr.15: 52581333-52581333 on Build GRCh38
rs868043820 Chr.15: 52581680-52581680 on Build GRCh38
rs142283183 Chr.15: 52581290-52581290 on Build GRCh38
rs903140628 Chr.15: 52581448-52581448 on Build GRCh38
rs1018420086 Chr.15: 52581651-52581651 on Build GRCh38
rs116213216 Chr.15: 52581600-52581600 on Build GRCh38
rs543239919 Chr.15: 52581352-52581352 on Build GRCh38
rs918553530 Chr.15: 52581362-52581362 on Build GRCh38

View Full Product Details