Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 78165497-78165990 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C17orf99
Entrez Gene ID
100141515
Gene Name
chromosome 17 open reading frame 99
Gene Aliases
UNQ464
UniGene
Hs.633034
Gene Chromosome Location
Chr.17: 78146035-78166283 on Build GRCh38
Transcripts
NM_001163075.1 XM_011524152.2 XM_017023999.1 XM_017024000.1 XM_017024001.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
985038 c.719G>A p.R240H Chr.17: 78165967-78165967 on Build GRCh38
6532571 c.711G>C p.R237S Chr.17: 78165959-78165959 on Build GRCh38
5387449 c.654G>A p.K218K Chr.17: 78165902-78165902 on Build GRCh38
985039 c.721C>T p.R241C Chr.17: 78165969-78165969 on Build GRCh38
3820762 c.651G>A p.Q217Q Chr.17: 78165899-78165899 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs1047314298 Chr.17: 78165795-78165795 on Build GRCh38
rs974679359 Chr.17: 78165584-78165584 on Build GRCh38
rs774573571 Chr.17: 78165630-78165630 on Build GRCh38
rs930799391 Chr.17: 78165764-78165764 on Build GRCh38
rs572209142 Chr.17: 78165825-78165825 on Build GRCh38
rs992796067 Chr.17: 78165671-78165671 on Build GRCh38
rs922666333 Chr.17: 78165757-78165757 on Build GRCh38
rs867675890 Chr.17: 78165914-78165914 on Build GRCh38
rs1022585284 Chr.17: 78165618-78165618 on Build GRCh38
rs541596488 Chr.17: 78165525-78165525 on Build GRCh38
rs922729853 Chr.17: 78165949-78165949 on Build GRCh38
rs886997843 Chr.17: 78165577-78165577 on Build GRCh38
rs942677216 Chr.17: 78165735-78165735 on Build GRCh38
rs1012654512 Chr.17: 78165656-78165656 on Build GRCh38
rs529004055 Chr.17: 78165798-78165798 on Build GRCh38
rs369058704 Chr.17: 78165613-78165613 on Build GRCh38
rs539863450 Chr.17: 78165753-78165753 on Build GRCh38
rs142542068 Chr.17: 78165780-78165780 on Build GRCh38
rs755900978 Chr.17: 78165750-78165750 on Build GRCh38
rs72898226 Chr.17: 78165524-78165524 on Build GRCh38
rs924658772 Chr.17: 78165666-78165666 on Build GRCh38
rs868659118 Chr.17: 78165919-78165919 on Build GRCh38
rs982642714 Chr.17: 78165961-78165961 on Build GRCh38
rs989490654 Chr.17: 78165705-78165705 on Build GRCh38
rs1016515009 Chr.17: 78165595-78165595 on Build GRCh38
rs1038242184 Chr.17: 78165811-78165811 on Build GRCh38
rs116745316 Chr.17: 78165832-78165832 on Build GRCh38
rs146249102 Chr.17: 78165702-78165702 on Build GRCh38
rs964068815 Chr.17: 78165908-78165908 on Build GRCh38
rs370940735 Chr.17: 78165843-78165843 on Build GRCh38
rs894236633 Chr.17: 78165632-78165632 on Build GRCh38
rs750873362 Chr.17: 78165920-78165920 on Build GRCh38
rs778329728 Chr.17: 78165794-78165794 on Build GRCh38
rs967888126 Chr.17: 78165646-78165646 on Build GRCh38
rs1031770878 Chr.17: 78165859-78165859 on Build GRCh38
rs759038653 Chr.17: 78165938-78165938 on Build GRCh38
rs762044423 Chr.17: 78165885-78165885 on Build GRCh38
rs555145060 Chr.17: 78165571-78165571 on Build GRCh38
rs111522426 Chr.17: 78165727-78165727 on Build GRCh38
rs200942539 Chr.17: 78165948-78165948 on Build GRCh38
rs1022538805 Chr.17: 78165903-78165903 on Build GRCh38
rs982672992 Chr.17: 78165662-78165662 on Build GRCh38
rs372937776 Chr.17: 78165966-78165966 on Build GRCh38
rs144746654 Chr.17: 78165826-78165826 on Build GRCh38
rs886149386 Chr.17: 78165786-78165786 on Build GRCh38
rs934653060 Chr.17: 78165668-78165668 on Build GRCh38
rs564892227 Chr.17: 78165812-78165812 on Build GRCh38
rs181766377 Chr.17: 78165888-78165888 on Build GRCh38
rs941247138 Chr.17: 78165800-78165800 on Build GRCh38
rs764333875 Chr.17: 78165590-78165590 on Build GRCh38
rs9906000 Chr.17: 78165659-78165659 on Build GRCh38
rs866154332 Chr.17: 78165797-78165797 on Build GRCh38
rs868781811 Chr.17: 78165600-78165600 on Build GRCh38
rs553113603 Chr.17: 78165950-78165950 on Build GRCh38
rs1018819879 Chr.17: 78165561-78165561 on Build GRCh38
rs955605505 Chr.17: 78165956-78165956 on Build GRCh38
rs545936530 Chr.17: 78165774-78165774 on Build GRCh38
rs1024471943 Chr.17: 78165658-78165658 on Build GRCh38
rs777325907 Chr.17: 78165838-78165838 on Build GRCh38
rs62079089 Chr.17: 78165682-78165682 on Build GRCh38

View Full Product Details