Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38296327-38296874 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5491355 c.1679G>A p.R560H Chr.2: 38296544-38296544 on Build GRCh38
5491356 c.1640G>A p.R547H Chr.2: 38296544-38296544 on Build GRCh38
1327090 c.1667G>C p.R556P Chr.2: 38296556-38296556 on Build GRCh38
5426831 c.1627C>T p.R543* Chr.2: 38296557-38296557 on Build GRCh38
5719580 c.1732A>C p.K578Q Chr.2: 38296491-38296491 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs773741290 Chr.2: 38296599-38296599 on Build GRCh38
rs555721463 Chr.2: 38296617-38296617 on Build GRCh38
rs773871722 Chr.2: 38296438-38296438 on Build GRCh38
rs778587117 Chr.2: 38296563-38296563 on Build GRCh38
rs1006847396 Chr.2: 38296798-38296798 on Build GRCh38
rs777729812 Chr.2: 38296642-38296642 on Build GRCh38
rs918646374 Chr.2: 38296674-38296674 on Build GRCh38
rs779451260 Chr.2: 38296750-38296750 on Build GRCh38
rs753753910 Chr.2: 38296659-38296659 on Build GRCh38
rs1024181083 Chr.2: 38296672-38296672 on Build GRCh38
rs995626514 Chr.2: 38296581-38296581 on Build GRCh38
rs917159852 Chr.2: 38296397-38296397 on Build GRCh38
rs773641783 Chr.2: 38296446-38296446 on Build GRCh38
rs138892566 Chr.2: 38296540-38296540 on Build GRCh38
rs781332270 Chr.2: 38296545-38296545 on Build GRCh38
rs775295361 Chr.2: 38296358-38296359 on Build GRCh38
rs777072430 Chr.2: 38296469-38296469 on Build GRCh38
rs767508888 Chr.2: 38296523-38296524 on Build GRCh38
rs763637458 Chr.2: 38296828-38296828 on Build GRCh38
rs1031275868 Chr.2: 38296775-38296775 on Build GRCh38
rs375283403 Chr.2: 38296605-38296605 on Build GRCh38
rs757629372 Chr.2: 38296556-38296556 on Build GRCh38
rs745510379 Chr.2: 38296745-38296745 on Build GRCh38
rs763419965 Chr.2: 38296454-38296457 on Build GRCh38
rs778764198 Chr.2: 38296709-38296709 on Build GRCh38
rs977018424 Chr.2: 38296413-38296413 on Build GRCh38
rs770529152 Chr.2: 38296437-38296437 on Build GRCh38
rs752776178 Chr.2: 38296635-38296635 on Build GRCh38
rs921467451 Chr.2: 38296458-38296458 on Build GRCh38
rs771547878 Chr.2: 38296573-38296573 on Build GRCh38
rs368318436 Chr.2: 38296754-38296754 on Build GRCh38
rs765700671 Chr.2: 38296501-38296501 on Build GRCh38
rs745520988 Chr.2: 38296568-38296568 on Build GRCh38
rs750908226 Chr.2: 38296727-38296729 on Build GRCh38
rs766834125 Chr.2: 38296476-38296477 on Build GRCh38
rs570815757 Chr.2: 38296405-38296405 on Build GRCh38
rs770373543 Chr.2: 38296594-38296594 on Build GRCh38
rs757101951 Chr.2: 38296678-38296678 on Build GRCh38
rs958764980 Chr.2: 38296354-38296354 on Build GRCh38
rs750757216 Chr.2: 38296508-38296508 on Build GRCh38
rs752280172 Chr.2: 38296715-38296715 on Build GRCh38
rs541492278 Chr.2: 38296813-38296813 on Build GRCh38
rs767021488 Chr.2: 38296449-38296449 on Build GRCh38
rs766666023 Chr.2: 38296539-38296539 on Build GRCh38
rs758714389 Chr.2: 38296523-38296523 on Build GRCh38
rs767735423 Chr.2: 38296648-38296648 on Build GRCh38
rs762376838 Chr.2: 38296470-38296470 on Build GRCh38
rs774439474 Chr.2: 38296637-38296638 on Build GRCh38
rs1048191826 Chr.2: 38296767-38296767 on Build GRCh38
rs766824661 Chr.2: 38296608-38296608 on Build GRCh38
rs759015618 Chr.2: 38296442-38296442 on Build GRCh38
rs912831076 Chr.2: 38296823-38296823 on Build GRCh38
rs1051385674 Chr.2: 38296497-38296497 on Build GRCh38
rs772930749 Chr.2: 38296434-38296434 on Build GRCh38
rs924373914 Chr.2: 38296414-38296414 on Build GRCh38
rs771697199 Chr.2: 38296747-38296747 on Build GRCh38
rs748941083 Chr.2: 38296435-38296435 on Build GRCh38
rs932837577 Chr.2: 38296797-38296797 on Build GRCh38
rs993931703 Chr.2: 38296438-38296439 on Build GRCh38
rs755058555 Chr.2: 38296544-38296544 on Build GRCh38
rs772564925 Chr.2: 38296712-38296713 on Build GRCh38
rs1057156723 Chr.2: 38296650-38296650 on Build GRCh38
rs891343712 Chr.2: 38296524-38296524 on Build GRCh38
rs958597454 Chr.2: 38296627-38296627 on Build GRCh38
rs762238785 Chr.2: 38296579-38296579 on Build GRCh38
rs754118937 Chr.2: 38296613-38296615 on Build GRCh38
rs932800095 Chr.2: 38296472-38296472 on Build GRCh38
rs770682788 Chr.2: 38296560-38296560 on Build GRCh38
rs374739524 Chr.2: 38296479-38296479 on Build GRCh38
rs989945345 Chr.2: 38296779-38296779 on Build GRCh38
rs759557987 Chr.2: 38296642-38296643 on Build GRCh38
rs774997459 Chr.2: 38296576-38296576 on Build GRCh38
rs997959872 Chr.2: 38296510-38296510 on Build GRCh38
rs139875817 Chr.2: 38296819-38296819 on Build GRCh38
rs781713137 Chr.2: 38296611-38296611 on Build GRCh38
rs376616532 Chr.2: 38296557-38296557 on Build GRCh38
rs527897508 Chr.2: 38296667-38296667 on Build GRCh38
rs767512084 Chr.2: 38296626-38296626 on Build GRCh38
rs370349486 Chr.2: 38296559-38296559 on Build GRCh38
rs755780114 Chr.2: 38296805-38296805 on Build GRCh38
rs765046929 Chr.2: 38296741-38296742 on Build GRCh38
rs866599237 Chr.2: 38296586-38296586 on Build GRCh38
rs1030744977 Chr.2: 38296515-38296515 on Build GRCh38
rs192994030 Chr.2: 38296636-38296636 on Build GRCh38
rs868275641 Chr.2: 38296476-38296476 on Build GRCh38
rs748044193 Chr.2: 38296554-38296554 on Build GRCh38
rs921005894 Chr.2: 38296785-38296785 on Build GRCh38
rs957244302 Chr.2: 38296778-38296778 on Build GRCh38
rs767314377 Chr.2: 38296383-38296383 on Build GRCh38
rs977071240 Chr.2: 38296758-38296758 on Build GRCh38

View Full Product Details