Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 38310118-38310615 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATL2
Entrez Gene ID
64225
Gene Name
atlastin GTPase 2
Gene Aliases
ARL3IP2, ARL6IP2, atlastin2
UniGene
Hs.594950
Gene Chromosome Location
Chr.2: 38293957-38378579 on Build GRCh38
Transcripts
NM_001135673.2 NM_001308076.1 NM_022374.3 NR_024191.1 XM_011533017.1 XM_011533018.2 XM_011533019.2 XM_011533020.1 XM_011533022.2 XM_011533023.2 XM_017004686.1 XM_017004687.1 XM_017004688.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7283651 c.751-9C>T p.? Chr.2: 38310456-38310456 on Build GRCh38
3839502 c.932G>A p.G311E Chr.2: 38310320-38310320 on Build GRCh38
5652421 c.934A>G p.R312G Chr.2: 38310318-38310318 on Build GRCh38
5829460 c.761A>G p.N254S Chr.2: 38310437-38310437 on Build GRCh38
4821708 c.874G>C p.G292R Chr.2: 38310378-38310378 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs537577400 Chr.2: 38310562-38310562 on Build GRCh38
rs555979551 Chr.2: 38310573-38310573 on Build GRCh38
rs377597782 Chr.2: 38310273-38310273 on Build GRCh38
rs112531332 Chr.2: 38310255-38310255 on Build GRCh38
rs138223839 Chr.2: 38310339-38310339 on Build GRCh38
rs574364224 Chr.2: 38310235-38310235 on Build GRCh38
rs187003240 Chr.2: 38310277-38310277 on Build GRCh38
rs377702950 Chr.2: 38310430-38310430 on Build GRCh38
rs925491208 Chr.2: 38310233-38310233 on Build GRCh38
rs371141139 Chr.2: 38310456-38310456 on Build GRCh38
rs953168805 Chr.2: 38310172-38310172 on Build GRCh38
rs778390170 Chr.2: 38310283-38310283 on Build GRCh38
rs150956803 Chr.2: 38310333-38310333 on Build GRCh38
rs919066207 Chr.2: 38310570-38310570 on Build GRCh38
rs898360630 Chr.2: 38310395-38310395 on Build GRCh38
rs780730002 Chr.2: 38310462-38310465 on Build GRCh38
rs6718249 Chr.2: 38310474-38310474 on Build GRCh38
rs780504335 Chr.2: 38310377-38310377 on Build GRCh38
rs754575751 Chr.2: 38310284-38310284 on Build GRCh38
rs747225826 Chr.2: 38310414-38310414 on Build GRCh38
rs551791012 Chr.2: 38310528-38310528 on Build GRCh38
rs746341011 Chr.2: 38310330-38310330 on Build GRCh38
rs754386802 Chr.2: 38310383-38310383 on Build GRCh38
rs752113211 Chr.2: 38310481-38310481 on Build GRCh38
rs565692584 Chr.2: 38310510-38310510 on Build GRCh38
rs762723091 Chr.2: 38310348-38310348 on Build GRCh38
rs2305245 Chr.2: 38310484-38310484 on Build GRCh38
rs775659596 Chr.2: 38310317-38310317 on Build GRCh38
rs757660514 Chr.2: 38310384-38310384 on Build GRCh38
rs965811790 Chr.2: 38310404-38310404 on Build GRCh38
rs189445621 Chr.2: 38310239-38310239 on Build GRCh38
rs966868339 Chr.2: 38310197-38310197 on Build GRCh38
rs1037781965 Chr.2: 38310386-38310386 on Build GRCh38
rs768709866 Chr.2: 38310323-38310323 on Build GRCh38
rs533184330 Chr.2: 38310522-38310522 on Build GRCh38
rs779728931 Chr.2: 38310276-38310276 on Build GRCh38
rs149189423 Chr.2: 38310189-38310189 on Build GRCh38
rs760794817 Chr.2: 38310319-38310319 on Build GRCh38
rs754504385 Chr.2: 38310280-38310280 on Build GRCh38
rs948120826 Chr.2: 38310270-38310270 on Build GRCh38
rs768783771 Chr.2: 38310418-38310418 on Build GRCh38
rs570046708 Chr.2: 38310552-38310552 on Build GRCh38
rs957760632 Chr.2: 38310561-38310561 on Build GRCh38
rs772242400 Chr.2: 38310297-38310297 on Build GRCh38
rs959048587 Chr.2: 38310187-38310187 on Build GRCh38
rs772405223 Chr.2: 38310407-38310407 on Build GRCh38
rs755618538 Chr.2: 38310483-38310483 on Build GRCh38
rs72803183 Chr.2: 38310185-38310185 on Build GRCh38
rs201184057 Chr.2: 38310439-38310439 on Build GRCh38
rs758829604 Chr.2: 38310496-38310496 on Build GRCh38
rs766165440 Chr.2: 38310361-38310361 on Build GRCh38
rs35287539 Chr.2: 38310454-38310454 on Build GRCh38
rs917560567 Chr.2: 38310194-38310194 on Build GRCh38
rs373365211 Chr.2: 38310399-38310399 on Build GRCh38
rs749758095 Chr.2: 38310284-38310284 on Build GRCh38
rs1015844510 Chr.2: 38310152-38310152 on Build GRCh38
rs995321434 Chr.2: 38310427-38310427 on Build GRCh38
rs767080650 Chr.2: 38310475-38310475 on Build GRCh38
rs372883231 Chr.2: 38310258-38310258 on Build GRCh38
rs776696907 Chr.2: 38310329-38310329 on Build GRCh38
rs746271599 Chr.2: 38310406-38310406 on Build GRCh38
rs778454626 Chr.2: 38310234-38310234 on Build GRCh38
rs201917247 Chr.2: 38310403-38310403 on Build GRCh38
rs780327810 Chr.2: 38310497-38310497 on Build GRCh38
rs780313529 Chr.2: 38310412-38310412 on Build GRCh38
rs771333710 Chr.2: 38310287-38310287 on Build GRCh38
rs2305244 Chr.2: 38310154-38310154 on Build GRCh38
rs34873284 Chr.2: 38310437-38310437 on Build GRCh38
rs755884524 Chr.2: 38310149-38310149 on Build GRCh38
rs995455913 Chr.2: 38310156-38310156 on Build GRCh38
rs143321385 Chr.2: 38310214-38310214 on Build GRCh38
rs1032036200 Chr.2: 38310523-38310523 on Build GRCh38
rs370235244 Chr.2: 38310289-38310289 on Build GRCh38
rs1005415361 Chr.2: 38310381-38310381 on Build GRCh38
rs61758717 Chr.2: 38310373-38310373 on Build GRCh38
rs765093782 Chr.2: 38310332-38310332 on Build GRCh38
rs761831630 Chr.2: 38310434-38310434 on Build GRCh38
rs558697052 Chr.2: 38310582-38310582 on Build GRCh38
rs1036177986 Chr.2: 38310316-38310316 on Build GRCh38
rs190242889 Chr.2: 38310486-38310486 on Build GRCh38
rs965905243 Chr.2: 38310212-38310212 on Build GRCh38
rs762887411 Chr.2: 38310452-38310452 on Build GRCh38
rs751231784 Chr.2: 38310370-38310370 on Build GRCh38
rs1031159979 Chr.2: 38310455-38310455 on Build GRCh38
rs111231844 Chr.2: 38310462-38310462 on Build GRCh38
rs773248420 Chr.2: 38310440-38310440 on Build GRCh38
rs751264585 Chr.2: 38310461-38310461 on Build GRCh38

View Full Product Details