Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 96646294-96646799 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FER1L5
Entrez Gene ID
90342
Gene Name
fer-1 like family member 5
Gene Aliases
-
UniGene
Hs.534025
Gene Chromosome Location
Chr.2: 96642719-96704887 on Build GRCh38
Transcripts
NM_001293083.1 XM_011512110.2 XM_011512111.2 XM_011512113.2 XM_011512114.2 XM_011512115.2 XM_011512116.2 XM_011512117.2 XM_011512118.2 XM_011512119.2 XM_011512120.2 XM_011512122.2 XM_011512125.2 XM_011512126.2 XM_011512127.2 XM_011512128.2 XM_017005222.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4913284 c.130T>A p.W44R Chr.2: 96646445-96646445 on Build GRCh38
1023935 c.93G>T p.K31N Chr.2: 96646408-96646408 on Build GRCh38
6046735 c.127G>A p.V43M Chr.2: 96646442-96646442 on Build GRCh38
4913285 c.12T>A p.C4* Chr.2: 96646445-96646445 on Build GRCh38
6046736 c.9G>A p.P3P Chr.2: 96646442-96646442 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs969798346 Chr.2: 96646340-96646340 on Build GRCh38
rs186957094 Chr.2: 96646318-96646318 on Build GRCh38
rs776697779 Chr.2: 96646486-96646488 on Build GRCh38
rs933893005 Chr.2: 96646418-96646418 on Build GRCh38
rs748400741 Chr.2: 96646584-96646584 on Build GRCh38
rs552314104 Chr.2: 96646627-96646627 on Build GRCh38
rs892062225 Chr.2: 96646524-96646524 on Build GRCh38
rs532053300 Chr.2: 96646646-96646646 on Build GRCh38
rs1044038900 Chr.2: 96646703-96646703 on Build GRCh38
rs764234083 Chr.2: 96646464-96646464 on Build GRCh38
rs769487296 Chr.2: 96646617-96646617 on Build GRCh38
rs761924602 Chr.2: 96646528-96646528 on Build GRCh38
rs913365797 Chr.2: 96646745-96646745 on Build GRCh38
rs1016717692 Chr.2: 96646616-96646616 on Build GRCh38
rs368841036 Chr.2: 96646442-96646442 on Build GRCh38
rs974695426 Chr.2: 96646676-96646676 on Build GRCh38
rs767171458 Chr.2: 96646389-96646389 on Build GRCh38
rs1056639356 Chr.2: 96646446-96646446 on Build GRCh38
rs930377974 Chr.2: 96646338-96646338 on Build GRCh38
rs532606570 Chr.2: 96646568-96646568 on Build GRCh38
rs754135057 Chr.2: 96646410-96646410 on Build GRCh38
rs374710689 Chr.2: 96646424-96646424 on Build GRCh38
rs1014351414 Chr.2: 96646321-96646321 on Build GRCh38
rs576406008 Chr.2: 96646582-96646582 on Build GRCh38
rs373982838 Chr.2: 96646497-96646497 on Build GRCh38
rs538191596 Chr.2: 96646679-96646679 on Build GRCh38
rs747041075 Chr.2: 96646352-96646352 on Build GRCh38
rs920290917 Chr.2: 96646322-96646322 on Build GRCh38
rs762906045 Chr.2: 96646774-96646774 on Build GRCh38
rs767726129 Chr.2: 96646682-96646682 on Build GRCh38
rs1028445646 Chr.2: 96646569-96646569 on Build GRCh38
rs1014469642 Chr.2: 96646538-96646538 on Build GRCh38
rs750632669 Chr.2: 96646726-96646726 on Build GRCh38
rs939134845 Chr.2: 96646670-96646670 on Build GRCh38
rs71427088 Chr.2: 96646413-96646413 on Build GRCh38
rs1009833682 Chr.2: 96646503-96646503 on Build GRCh38
rs1023950786 Chr.2: 96646329-96646329 on Build GRCh38
rs773795581 Chr.2: 96646516-96646516 on Build GRCh38
rs1001200528 Chr.2: 96646614-96646614 on Build GRCh38
rs1039127722 Chr.2: 96646471-96646471 on Build GRCh38
rs943464174 Chr.2: 96646450-96646450 on Build GRCh38
rs955470861 Chr.2: 96646647-96646647 on Build GRCh38
rs936077456 Chr.2: 96646683-96646683 on Build GRCh38
rs765614142 Chr.2: 96646441-96646441 on Build GRCh38
rs575151063 Chr.2: 96646692-96646692 on Build GRCh38
rs986700422 Chr.2: 96646663-96646663 on Build GRCh38
rs918526287 Chr.2: 96646396-96646396 on Build GRCh38
rs926336954 Chr.2: 96646666-96646666 on Build GRCh38
rs1043731702 Chr.2: 96646543-96646543 on Build GRCh38
rs904613468 Chr.2: 96646491-96646491 on Build GRCh38
rs141863954 Chr.2: 96646758-96646758 on Build GRCh38
rs911129376 Chr.2: 96646669-96646669 on Build GRCh38
rs757483840 Chr.2: 96646499-96646499 on Build GRCh38
rs980415122 Chr.2: 96646654-96646654 on Build GRCh38
rs962257361 Chr.2: 96646633-96646633 on Build GRCh38
rs569231901 Chr.2: 96646638-96646638 on Build GRCh38
rs1025450459 Chr.2: 96646631-96646631 on Build GRCh38
rs972989333 Chr.2: 96646630-96646630 on Build GRCh38
rs148056683 Chr.2: 96646755-96646755 on Build GRCh38
rs972575882 Chr.2: 96646381-96646381 on Build GRCh38
rs903845450 Chr.2: 96646707-96646707 on Build GRCh38
rs936195486 Chr.2: 96646494-96646494 on Build GRCh38
rs964720434 Chr.2: 96646675-96646675 on Build GRCh38
rs1016688080 Chr.2: 96646353-96646353 on Build GRCh38
rs962577522 Chr.2: 96646359-96646359 on Build GRCh38
rs972030317 Chr.2: 96646636-96646636 on Build GRCh38
rs914101856 Chr.2: 96646673-96646673 on Build GRCh38
rs776761351 Chr.2: 96646553-96646553 on Build GRCh38

View Full Product Details