Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 219385540-219386035 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNPEP
Entrez Gene ID
23549
Gene Name
aspartyl aminopeptidase
Gene Aliases
ASPEP, DAP
UniGene
Hs.258551
Gene Chromosome Location
Chr.2: 219372029-219400022 on Build GRCh38
Transcripts
NM_001319116.1 NM_001319117.1 NM_001319118.1 NM_001319119.1 NM_001319120.1 NM_001319121.1 NM_001319122.1 NM_012100.3 NR_134970.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1191915 c.652C>T p.P218S Chr.2: 219385645-219385645 on Build GRCh38
6647885 c.568G>T p.G190W Chr.2: 219385990-219385990 on Build GRCh38
4422403 c.577A>G p.T193A Chr.2: 219385981-219385981 on Build GRCh38
292997 c.559G>A p.E187K Chr.2: 219385999-219385999 on Build GRCh38
7413456 c.643G>T p.E215* Chr.2: 219385654-219385654 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs772039443 Chr.2: 219385635-219385635 on Build GRCh38
rs572324192 Chr.2: 219385830-219385830 on Build GRCh38
rs200325612 Chr.2: 219385738-219385738 on Build GRCh38
rs961906436 Chr.2: 219385699-219385699 on Build GRCh38
rs996523409 Chr.2: 219385646-219385646 on Build GRCh38
rs915815789 Chr.2: 219385847-219385847 on Build GRCh38
rs757131932 Chr.2: 219385999-219385999 on Build GRCh38
rs758143872 Chr.2: 219385628-219385629 on Build GRCh38
rs762195603 Chr.2: 219385745-219385745 on Build GRCh38
rs751923522 Chr.2: 219385692-219385692 on Build GRCh38
rs991668448 Chr.2: 219385974-219385974 on Build GRCh38
rs968852572 Chr.2: 219386001-219386001 on Build GRCh38
rs72963541 Chr.2: 219385710-219385710 on Build GRCh38
rs760793094 Chr.2: 219385767-219385767 on Build GRCh38
rs1015390675 Chr.2: 219385834-219385834 on Build GRCh38
rs112114380 Chr.2: 219385649-219385649 on Build GRCh38
rs776258705 Chr.2: 219385919-219385919 on Build GRCh38
rs751831593 Chr.2: 219385593-219385593 on Build GRCh38
rs934862443 Chr.2: 219385777-219385777 on Build GRCh38
rs748003371 Chr.2: 219385633-219385633 on Build GRCh38
rs934727938 Chr.2: 219385887-219385887 on Build GRCh38
rs777876217 Chr.2: 219385917-219385917 on Build GRCh38
rs114643426 Chr.2: 219385820-219385820 on Build GRCh38
rs759720238 Chr.2: 219385981-219385981 on Build GRCh38
rs201369535 Chr.2: 219385638-219385638 on Build GRCh38
rs779696546 Chr.2: 219385944-219385944 on Build GRCh38
rs776090081 Chr.2: 219385655-219385655 on Build GRCh38
rs762285257 Chr.2: 219385956-219385956 on Build GRCh38
rs777788328 Chr.2: 219385720-219385720 on Build GRCh38
rs1029277934 Chr.2: 219385610-219385610 on Build GRCh38
rs1039202004 Chr.2: 219385925-219385925 on Build GRCh38
rs778939895 Chr.2: 219385628-219385628 on Build GRCh38
rs758407040 Chr.2: 219385719-219385719 on Build GRCh38
rs774878526 Chr.2: 219385950-219385950 on Build GRCh38
rs201832630 Chr.2: 219385631-219385631 on Build GRCh38
rs546932625 Chr.2: 219385823-219385823 on Build GRCh38
rs566757039 Chr.2: 219385883-219385883 on Build GRCh38
rs370821247 Chr.2: 219385946-219385946 on Build GRCh38
rs774786551 Chr.2: 219385744-219385744 on Build GRCh38
rs750571417 Chr.2: 219385712-219385712 on Build GRCh38
rs865842460 Chr.2: 219385686-219385686 on Build GRCh38
rs767415848 Chr.2: 219386008-219386008 on Build GRCh38
rs757555610 Chr.2: 219385705-219385705 on Build GRCh38
rs759112997 Chr.2: 219385657-219385657 on Build GRCh38
rs779600424 Chr.2: 219386015-219386015 on Build GRCh38
rs376611555 Chr.2: 219385599-219385599 on Build GRCh38
rs754065059 Chr.2: 219385973-219385973 on Build GRCh38
rs761036269 Chr.2: 219385960-219385960 on Build GRCh38
rs372778549 Chr.2: 219385570-219385570 on Build GRCh38
rs555789796 Chr.2: 219385882-219385882 on Build GRCh38
rs978754957 Chr.2: 219385868-219385868 on Build GRCh38
rs772543358 Chr.2: 219385957-219385957 on Build GRCh38
rs541417051 Chr.2: 219385838-219385838 on Build GRCh38
rs768541208 Chr.2: 219385708-219385709 on Build GRCh38
rs376997345 Chr.2: 219385991-219385991 on Build GRCh38
rs750705119 Chr.2: 219385760-219385760 on Build GRCh38
rs779725870 Chr.2: 219385630-219385630 on Build GRCh38
rs745594523 Chr.2: 219385932-219385932 on Build GRCh38
rs773443381 Chr.2: 219385958-219385958 on Build GRCh38
rs900762385 Chr.2: 219385927-219385927 on Build GRCh38
rs769215402 Chr.2: 219386011-219386011 on Build GRCh38
rs766549206 Chr.2: 219385769-219385769 on Build GRCh38
rs535999154 Chr.2: 219385739-219385739 on Build GRCh38
rs908406667 Chr.2: 219385704-219385704 on Build GRCh38
rs745970030 Chr.2: 219385607-219385607 on Build GRCh38
rs750196242 Chr.2: 219386010-219386010 on Build GRCh38
rs533999406 Chr.2: 219385941-219385941 on Build GRCh38
rs193041992 Chr.2: 219385731-219385731 on Build GRCh38
rs766524271 Chr.2: 219385968-219385968 on Build GRCh38
rs374562682 Chr.2: 219385902-219385902 on Build GRCh38
rs751505940 Chr.2: 219385918-219385918 on Build GRCh38
rs780934041 Chr.2: 219385730-219385730 on Build GRCh38
rs145015119 Chr.2: 219385718-219385718 on Build GRCh38
rs774852618 Chr.2: 219385735-219385735 on Build GRCh38
rs764507131 Chr.2: 219385688-219385688 on Build GRCh38
rs769333907 Chr.2: 219385943-219385943 on Build GRCh38
rs186702343 Chr.2: 219385618-219385618 on Build GRCh38
rs763431791 Chr.2: 219385687-219385687 on Build GRCh38
rs370933655 Chr.2: 219385716-219385716 on Build GRCh38
rs764414917 Chr.2: 219385582-219385582 on Build GRCh38
rs764616876 Chr.2: 219385665-219385665 on Build GRCh38
rs149080839 Chr.2: 219385728-219385728 on Build GRCh38
rs561003440 Chr.2: 219385888-219385888 on Build GRCh38
rs751600279 Chr.2: 219385995-219385995 on Build GRCh38
rs753869174 Chr.2: 219385794-219385794 on Build GRCh38
rs915648027 Chr.2: 219385992-219385992 on Build GRCh38
rs993339738 Chr.2: 219385945-219385945 on Build GRCh38
rs755051247 Chr.2: 219385797-219385797 on Build GRCh38
rs992712622 Chr.2: 219385839-219385839 on Build GRCh38
rs377452687 Chr.2: 219385574-219385574 on Build GRCh38
rs746726545 Chr.2: 219385639-219385639 on Build GRCh38
rs201097301 Chr.2: 219385916-219385916 on Build GRCh38
rs1009596755 Chr.2: 219385604-219385604 on Build GRCh38
rs878437 Chr.2: 219385600-219385600 on Build GRCh38
rs746949766 Chr.2: 219385692-219385693 on Build GRCh38
rs778294181 Chr.2: 219385569-219385569 on Build GRCh38
rs776107961 Chr.2: 219385678-219385678 on Build GRCh38
rs767741365 Chr.2: 219385759-219385759 on Build GRCh38
rs969986725 Chr.2: 219385855-219385855 on Build GRCh38
rs780829752 Chr.2: 219385930-219385930 on Build GRCh38
rs1041076731 Chr.2: 219385796-219385796 on Build GRCh38

View Full Product Details