Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46354931-46355425 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528831.1 XM_011528832.1 XM_011528833.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_011528837.1 XM_011528838.1 XM_017027856.1 XM_017027857.1 XM_017027858.1 XM_017027859.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6781671 c.617G>A p.G206D Chr.20: 46355120-46355120 on Build GRCh38
6858836 c.525C>T p.I175I Chr.20: 46355125-46355125 on Build GRCh38
4547080 c.15G>A p.A5A Chr.20: 46355236-46355236 on Build GRCh38
5462968 c.634A>G p.T212A Chr.20: 46355103-46355103 on Build GRCh38
7208801 c.586A>T p.N196Y Chr.20: 46354957-46354957 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs112467299 Chr.20: 46354967-46354967 on Build GRCh38
rs752180989 Chr.20: 46355068-46355068 on Build GRCh38
rs760485714 Chr.20: 46355317-46355317 on Build GRCh38
rs764906181 Chr.20: 46355383-46355383 on Build GRCh38
rs775288921 Chr.20: 46355235-46355235 on Build GRCh38
rs140165611 Chr.20: 46355061-46355061 on Build GRCh38
rs771350617 Chr.20: 46355097-46355097 on Build GRCh38
rs752731371 Chr.20: 46355163-46355163 on Build GRCh38
rs746325623 Chr.20: 46355099-46355099 on Build GRCh38
rs767328554 Chr.20: 46355152-46355152 on Build GRCh38
rs776755894 Chr.20: 46355337-46355337 on Build GRCh38
rs753479598 Chr.20: 46355181-46355181 on Build GRCh38
rs5841635 Chr.20: 46355368-46355368 on Build GRCh38
rs576707210 Chr.20: 46355014-46355014 on Build GRCh38
rs752892811 Chr.20: 46354975-46354975 on Build GRCh38
rs925562333 Chr.20: 46355156-46355156 on Build GRCh38
rs760670631 Chr.20: 46355236-46355236 on Build GRCh38
rs1012797679 Chr.20: 46355020-46355020 on Build GRCh38
rs377660747 Chr.20: 46355239-46355239 on Build GRCh38
rs373176914 Chr.20: 46354987-46354987 on Build GRCh38
rs771701215 Chr.20: 46355283-46355283 on Build GRCh38
rs773465080 Chr.20: 46355230-46355230 on Build GRCh38
rs377754175 Chr.20: 46355234-46355234 on Build GRCh38
rs772484930 Chr.20: 46355115-46355115 on Build GRCh38
rs751379631 Chr.20: 46355140-46355140 on Build GRCh38
rs943635006 Chr.20: 46355151-46355151 on Build GRCh38
rs149565718 Chr.20: 46354964-46354964 on Build GRCh38
rs1055461893 Chr.20: 46355272-46355272 on Build GRCh38
rs749011443 Chr.20: 46355093-46355093 on Build GRCh38
rs555641771 Chr.20: 46355397-46355397 on Build GRCh38
rs750214603 Chr.20: 46354988-46354988 on Build GRCh38
rs780415956 Chr.20: 46355201-46355201 on Build GRCh38
rs561925431 Chr.20: 46355205-46355205 on Build GRCh38
rs755570119 Chr.20: 46355164-46355164 on Build GRCh38
rs775373969 Chr.20: 46355316-46355316 on Build GRCh38
rs780573785 Chr.20: 46355006-46355006 on Build GRCh38
rs201253430 Chr.20: 46354998-46354998 on Build GRCh38
rs761392892 Chr.20: 46355354-46355354 on Build GRCh38
rs371537544 Chr.20: 46355066-46355066 on Build GRCh38
rs779944841 Chr.20: 46355000-46355000 on Build GRCh38
rs781210971 Chr.20: 46355267-46355267 on Build GRCh38
rs776705741 Chr.20: 46355209-46355209 on Build GRCh38
rs756988407 Chr.20: 46355183-46355183 on Build GRCh38
rs200170864 Chr.20: 46355277-46355277 on Build GRCh38
rs761975946 Chr.20: 46355215-46355215 on Build GRCh38
rs187394710 Chr.20: 46355241-46355241 on Build GRCh38
rs34405647 Chr.20: 46355074-46355074 on Build GRCh38
rs760648850 Chr.20: 46355125-46355125 on Build GRCh38
rs764189719 Chr.20: 46355052-46355052 on Build GRCh38
rs200754558 Chr.20: 46355034-46355034 on Build GRCh38
rs777413234 Chr.20: 46355167-46355167 on Build GRCh38
rs754137411 Chr.20: 46355058-46355058 on Build GRCh38
rs963154012 Chr.20: 46355044-46355044 on Build GRCh38
rs540107970 Chr.20: 46355008-46355008 on Build GRCh38
rs766121379 Chr.20: 46355257-46355257 on Build GRCh38
rs764088747 Chr.20: 46355322-46355322 on Build GRCh38
rs759431389 Chr.20: 46355142-46355142 on Build GRCh38
rs192544192 Chr.20: 46355273-46355273 on Build GRCh38
rs775452102 Chr.20: 46355047-46355047 on Build GRCh38
rs756442379 Chr.20: 46355086-46355086 on Build GRCh38
rs749588369 Chr.20: 46355022-46355022 on Build GRCh38
rs749594373 Chr.20: 46355282-46355282 on Build GRCh38
rs779429934 Chr.20: 46355098-46355098 on Build GRCh38
rs760908759 Chr.20: 46355048-46355048 on Build GRCh38
rs1025446513 Chr.20: 46355294-46355294 on Build GRCh38
rs151119784 Chr.20: 46355010-46355010 on Build GRCh38
rs776103624 Chr.20: 46355116-46355116 on Build GRCh38
rs202059286 Chr.20: 46355065-46355065 on Build GRCh38
rs868131865 Chr.20: 46355194-46355194 on Build GRCh38
rs768551340 Chr.20: 46355208-46355208 on Build GRCh38
rs775041650 Chr.20: 46354969-46354969 on Build GRCh38
rs368357747 Chr.20: 46355226-46355226 on Build GRCh38
rs370573129 Chr.20: 46355041-46355041 on Build GRCh38
rs537788869 Chr.20: 46355359-46355359 on Build GRCh38
rs759669175 Chr.20: 46354970-46354970 on Build GRCh38
rs776714228 Chr.20: 46355128-46355128 on Build GRCh38
rs371950415 Chr.20: 46355114-46355114 on Build GRCh38
rs537225524 Chr.20: 46355019-46355019 on Build GRCh38
rs759232565 Chr.20: 46355233-46355233 on Build GRCh38
rs768153291 Chr.20: 46355122-46355122 on Build GRCh38
rs201199146 Chr.20: 46355206-46355206 on Build GRCh38
rs764959329 Chr.20: 46355240-46355240 on Build GRCh38
rs555972774 Chr.20: 46355033-46355033 on Build GRCh38
rs79899554 Chr.20: 46355131-46355131 on Build GRCh38
rs753426739 Chr.20: 46355238-46355238 on Build GRCh38
rs370738121 Chr.20: 46355003-46355003 on Build GRCh38
rs199648810 Chr.20: 46354974-46354974 on Build GRCh38
rs547931791 Chr.20: 46355228-46355228 on Build GRCh38
rs756300684 Chr.20: 46354978-46354978 on Build GRCh38
rs748197102 Chr.20: 46355269-46355269 on Build GRCh38
rs573900074 Chr.20: 46355400-46355400 on Build GRCh38
rs779377683 Chr.20: 46355197-46355197 on Build GRCh38
rs746201204 Chr.20: 46355203-46355203 on Build GRCh38
rs531663384 Chr.20: 46355127-46355127 on Build GRCh38
rs199606072 Chr.20: 46355064-46355064 on Build GRCh38
rs5841636 Chr.20: 46355383-46355383 on Build GRCh38
rs758684785 Chr.20: 46355254-46355254 on Build GRCh38
rs763460018 Chr.20: 46355237-46355237 on Build GRCh38
rs770036106 Chr.20: 46355224-46355224 on Build GRCh38
rs755693821 Chr.20: 46355070-46355070 on Build GRCh38
rs747618033 Chr.20: 46355207-46355207 on Build GRCh38
rs755532746 Chr.20: 46355260-46355260 on Build GRCh38
rs747006886 Chr.20: 46355002-46355002 on Build GRCh38

View Full Product Details