Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 46364268-46364775 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SLC35C2
Entrez Gene ID
51006
Gene Name
solute carrier family 35 member C2
Gene Aliases
BA394O2.1, C20orf5, CGI-15, OVCOV1
UniGene
Hs.593344
Gene Chromosome Location
Chr.20: 46349528-46364461 on Build GRCh38
Transcripts
NM_001281457.1 NM_001281458.1 NM_001281459.1 NM_001281460.1 NM_015945.11 NM_173073.3 NM_173179.3 XM_011528832.1 XM_011528834.1 XM_011528835.1 XM_011528836.1 XM_017027857.1 XM_017027858.1 XM_017027860.1 XM_017027861.1 XM_017027862.1

SNP Detail

SNP ID Chromosome Location
rs979194229 Chr.20: 46364625-46364625 on Build GRCh38
rs139388788 Chr.20: 46364353-46364353 on Build GRCh38
rs868226435 Chr.20: 46364399-46364399 on Build GRCh38
rs987563084 Chr.20: 46364450-46364450 on Build GRCh38
rs1016813311 Chr.20: 46364342-46364342 on Build GRCh38
rs934626303 Chr.20: 46364655-46364655 on Build GRCh38
rs142135632 Chr.20: 46364636-46364636 on Build GRCh38
rs1005512576 Chr.20: 46364453-46364453 on Build GRCh38
rs149882367 Chr.20: 46364555-46364555 on Build GRCh38
rs1053754506 Chr.20: 46364501-46364502 on Build GRCh38
rs192333876 Chr.20: 46364612-46364612 on Build GRCh38
rs1014341131 Chr.20: 46364509-46364509 on Build GRCh38
rs546606483 Chr.20: 46364588-46364588 on Build GRCh38
rs540652472 Chr.20: 46364366-46364366 on Build GRCh38
rs910912060 Chr.20: 46364533-46364533 on Build GRCh38
rs148607706 Chr.20: 46364617-46364617 on Build GRCh38
rs904977162 Chr.20: 46364664-46364664 on Build GRCh38
rs1016863767 Chr.20: 46364464-46364464 on Build GRCh38
rs961199081 Chr.20: 46364344-46364344 on Build GRCh38
rs147797282 Chr.20: 46364654-46364654 on Build GRCh38
rs116254648 Chr.20: 46364554-46364554 on Build GRCh38
rs957082497 Chr.20: 46364708-46364708 on Build GRCh38
rs369101021 Chr.20: 46364297-46364297 on Build GRCh38
rs577506702 Chr.20: 46364435-46364435 on Build GRCh38
rs943388565 Chr.20: 46364474-46364474 on Build GRCh38
rs933727660 Chr.20: 46364356-46364356 on Build GRCh38
rs368813469 Chr.20: 46364419-46364419 on Build GRCh38
rs1051826005 Chr.20: 46364296-46364296 on Build GRCh38
rs1026203516 Chr.20: 46364565-46364565 on Build GRCh38
rs1007089772 Chr.20: 46364700-46364700 on Build GRCh38
rs1002307475 Chr.20: 46364541-46364541 on Build GRCh38
rs892237323 Chr.20: 46364348-46364348 on Build GRCh38
rs183652182 Chr.20: 46364652-46364652 on Build GRCh38
rs890369968 Chr.20: 46364723-46364723 on Build GRCh38
rs769140606 Chr.20: 46364536-46364536 on Build GRCh38
rs143857701 Chr.20: 46364576-46364576 on Build GRCh38
rs1053306363 Chr.20: 46364656-46364656 on Build GRCh38
rs910601649 Chr.20: 46364456-46364456 on Build GRCh38
rs371445419 Chr.20: 46364318-46364318 on Build GRCh38
rs376658352 Chr.20: 46364418-46364418 on Build GRCh38
rs985549572 Chr.20: 46364364-46364364 on Build GRCh38
rs563077173 Chr.20: 46364442-46364442 on Build GRCh38
rs533931286 Chr.20: 46364661-46364661 on Build GRCh38
rs936280862 Chr.20: 46364390-46364390 on Build GRCh38
rs1054627274 Chr.20: 46364398-46364398 on Build GRCh38
rs550323250 Chr.20: 46364624-46364624 on Build GRCh38
rs907293469 Chr.20: 46364426-46364426 on Build GRCh38
rs984017846 Chr.20: 46364335-46364335 on Build GRCh38
rs530579019 Chr.20: 46364503-46364503 on Build GRCh38
rs1024650531 Chr.20: 46364374-46364374 on Build GRCh38
rs946221441 Chr.20: 46364402-46364402 on Build GRCh38
rs1018024412 Chr.20: 46364705-46364705 on Build GRCh38
rs999463256 Chr.20: 46364333-46364333 on Build GRCh38
rs955228832 Chr.20: 46364695-46364695 on Build GRCh38
rs530846979 Chr.20: 46364638-46364638 on Build GRCh38
rs963314483 Chr.20: 46364420-46364420 on Build GRCh38
rs200869844 Chr.20: 46364524-46364524 on Build GRCh38
rs928800434 Chr.20: 46364556-46364556 on Build GRCh38
rs879657086 Chr.20: 46364363-46364363 on Build GRCh38
rs913492370 Chr.20: 46364545-46364545 on Build GRCh38
rs1029940288 Chr.20: 46364688-46364688 on Build GRCh38
rs972524588 Chr.20: 46364346-46364346 on Build GRCh38
rs1006234180 Chr.20: 46364635-46364635 on Build GRCh38
rs1026275259 Chr.20: 46364707-46364707 on Build GRCh38
rs904945899 Chr.20: 46364525-46364525 on Build GRCh38
rs887778316 Chr.20: 46364447-46364447 on Build GRCh38
rs955325461 Chr.20: 46364523-46364523 on Build GRCh38
rs957030708 Chr.20: 46364543-46364543 on Build GRCh38
rs994017379 Chr.20: 46364481-46364481 on Build GRCh38
rs989733450 Chr.20: 46364544-46364544 on Build GRCh38
rs994068194 Chr.20: 46364683-46364683 on Build GRCh38
rs934909583 Chr.20: 46364492-46364492 on Build GRCh38
rs374640158 Chr.20: 46364733-46364733 on Build GRCh38
rs910872011 Chr.20: 46364381-46364381 on Build GRCh38
rs544817940 Chr.20: 46364439-46364439 on Build GRCh38
rs961249573 Chr.20: 46364475-46364475 on Build GRCh38
rs897094700 Chr.20: 46364667-46364667 on Build GRCh38
rs910672233 Chr.20: 46364599-46364599 on Build GRCh38
rs917458186 Chr.20: 46364657-46364657 on Build GRCh38
rs1001876275 Chr.20: 46364405-46364405 on Build GRCh38
rs146394252 Chr.20: 46364319-46364319 on Build GRCh38
rs959568564 Chr.20: 46364324-46364324 on Build GRCh38
rs1001980853 Chr.20: 46364682-46364682 on Build GRCh38
rs1043703780 Chr.20: 46364321-46364321 on Build GRCh38

View Full Product Details