Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 32748310-32748823 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
PAXBP1
Entrez Gene ID
94104
Gene Name
PAX3 and PAX7 binding protein 1
Gene Aliases
BM020, C21orf66, FSAP105, GCFC, GCFC1
UniGene
Hs.644004
Gene Chromosome Location
Chr.21: 32733899-32771858 on Build GRCh38
Transcripts
NM_013329.3 NM_016631.3 NR_027873.1 XM_006724066.1 XM_011529804.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1030246 c.1787A>C p.D596A Chr.21: 32748635-32748635 on Build GRCh38
1208006 c.1843G>T p.D615Y Chr.21: 32748579-32748579 on Build GRCh38
5815979 c.1815G>A p.W605* Chr.21: 32748607-32748607 on Build GRCh38
4799722 c.1852A>G p.I618V Chr.21: 32748570-32748570 on Build GRCh38
6738310 c.1891C>T p.R631* Chr.21: 32748531-32748531 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374635092 Chr.21: 32748644-32748644 on Build GRCh38
rs774324834 Chr.21: 32748526-32748526 on Build GRCh38
rs766826727 Chr.21: 32748561-32748561 on Build GRCh38
rs759918754 Chr.21: 32748682-32748682 on Build GRCh38
rs759494657 Chr.21: 32748530-32748530 on Build GRCh38
rs370916423 Chr.21: 32748621-32748621 on Build GRCh38
rs992910295 Chr.21: 32748736-32748736 on Build GRCh38
rs375551594 Chr.21: 32748708-32748708 on Build GRCh38
rs1042731056 Chr.21: 32748485-32748485 on Build GRCh38
rs112853061 Chr.21: 32748678-32748678 on Build GRCh38
rs539334739 Chr.21: 32748744-32748744 on Build GRCh38
rs772015075 Chr.21: 32748620-32748620 on Build GRCh38
rs753526335 Chr.21: 32748551-32748551 on Build GRCh38
rs1009687330 Chr.21: 32748762-32748762 on Build GRCh38
rs983931768 Chr.21: 32748478-32748478 on Build GRCh38
rs769909362 Chr.21: 32748496-32748496 on Build GRCh38
rs748862473 Chr.21: 32748723-32748724 on Build GRCh38
rs373912940 Chr.21: 32748745-32748745 on Build GRCh38
rs140333561 Chr.21: 32748679-32748679 on Build GRCh38
rs143101864 Chr.21: 32748582-32748582 on Build GRCh38
rs191207306 Chr.21: 32748765-32748765 on Build GRCh38
rs958124653 Chr.21: 32748388-32748388 on Build GRCh38
rs1032406824 Chr.21: 32748347-32748347 on Build GRCh38
rs763712461 Chr.21: 32748546-32748546 on Build GRCh38
rs761093984 Chr.21: 32748476-32748476 on Build GRCh38
rs755573314 Chr.21: 32748570-32748570 on Build GRCh38
rs768272368 Chr.21: 32748748-32748748 on Build GRCh38
rs143526654 Chr.21: 32748520-32748520 on Build GRCh38
rs976661062 Chr.21: 32748438-32748438 on Build GRCh38
rs753240237 Chr.21: 32748695-32748695 on Build GRCh38
rs772982419 Chr.21: 32748747-32748747 on Build GRCh38
rs916948878 Chr.21: 32748591-32748591 on Build GRCh38
rs778713949 Chr.21: 32748605-32748605 on Build GRCh38
rs748218831 Chr.21: 32748455-32748455 on Build GRCh38
rs554459207 Chr.21: 32748409-32748409 on Build GRCh38
rs779897559 Chr.21: 32748734-32748734 on Build GRCh38
rs745720446 Chr.21: 32748730-32748730 on Build GRCh38
rs1005061193 Chr.21: 32748654-32748654 on Build GRCh38
rs113709185 Chr.21: 32748392-32748392 on Build GRCh38
rs763452858 Chr.21: 32748560-32748560 on Build GRCh38
rs371129196 Chr.21: 32748598-32748598 on Build GRCh38
rs958793637 Chr.21: 32748423-32748423 on Build GRCh38
rs988128736 Chr.21: 32748396-32748396 on Build GRCh38
rs368168645 Chr.21: 32748667-32748667 on Build GRCh38
rs564973557 Chr.21: 32748403-32748403 on Build GRCh38
rs552934902 Chr.21: 32748417-32748417 on Build GRCh38
rs992857908 Chr.21: 32748430-32748430 on Build GRCh38
rs774871105 Chr.21: 32748535-32748535 on Build GRCh38
rs746568370 Chr.21: 32748630-32748630 on Build GRCh38
rs757479207 Chr.21: 32748721-32748721 on Build GRCh38
rs777922716 Chr.21: 32748589-32748589 on Build GRCh38
rs963932139 Chr.21: 32748395-32748395 on Build GRCh38
rs187013524 Chr.21: 32748604-32748604 on Build GRCh38
rs779053341 Chr.21: 32748729-32748729 on Build GRCh38
rs370810041 Chr.21: 32748735-32748735 on Build GRCh38
rs760175703 Chr.21: 32748541-32748541 on Build GRCh38
rs1005586986 Chr.21: 32748352-32748352 on Build GRCh38
rs773652803 Chr.21: 32748440-32748440 on Build GRCh38
rs771961954 Chr.21: 32748534-32748534 on Build GRCh38
rs777989466 Chr.21: 32748499-32748499 on Build GRCh38
rs756201793 Chr.21: 32748587-32748587 on Build GRCh38
rs374674342 Chr.21: 32748602-32748602 on Build GRCh38
rs932475953 Chr.21: 32748477-32748477 on Build GRCh38
rs767964333 Chr.21: 32748690-32748690 on Build GRCh38
rs776309956 Chr.21: 32748739-32748739 on Build GRCh38
rs756186010 Chr.21: 32748701-32748701 on Build GRCh38
rs781675104 Chr.21: 32748579-32748579 on Build GRCh38
rs745599782 Chr.21: 32748614-32748614 on Build GRCh38
rs753878332 Chr.21: 32748720-32748720 on Build GRCh38
rs368191673 Chr.21: 32748731-32748731 on Build GRCh38
rs768182808 Chr.21: 32748639-32748639 on Build GRCh38
rs111524488 Chr.21: 32748725-32748725 on Build GRCh38
rs751922547 Chr.21: 32748569-32748569 on Build GRCh38
rs973380913 Chr.21: 32748406-32748406 on Build GRCh38
rs770705034 Chr.21: 32748523-32748523 on Build GRCh38
rs917770816 Chr.21: 32748437-32748437 on Build GRCh38
rs1015170132 Chr.21: 32748382-32748382 on Build GRCh38

View Full Product Details