Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 24541170-24541679 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GUCD1
Entrez Gene ID
83606
Gene Name
guanylyl cyclase domain containing 1
Gene Aliases
C22orf13, LLN4
UniGene
Hs.9850
Gene Chromosome Location
Chr.22: 24540423-24555935 on Build GRCh38
Transcripts
NM_001284251.1 NM_001284252.1 NM_001284253.1 NM_001284254.1 NM_001284255.1 NM_001284256.1 NM_001284257.1 NM_031444.3 NR_104286.1 XM_005261761.1

SNP Detail

SNP ID Chromosome Location
rs567333379 Chr.22: 24541365-24541365 on Build GRCh38
rs968700380 Chr.22: 24541494-24541494 on Build GRCh38
rs987343231 Chr.22: 24541628-24541628 on Build GRCh38
rs573171127 Chr.22: 24541514-24541514 on Build GRCh38
rs971908930 Chr.22: 24541380-24541380 on Build GRCh38
rs71660760 Chr.22: 24541629-24541630 on Build GRCh38
rs996709055 Chr.22: 24541644-24541644 on Build GRCh38
rs1016130203 Chr.22: 24541454-24541454 on Build GRCh38
rs553336307 Chr.22: 24541590-24541591 on Build GRCh38
rs999055442 Chr.22: 24541287-24541287 on Build GRCh38
rs552507514 Chr.22: 24541417-24541417 on Build GRCh38
rs534365067 Chr.22: 24541410-24541410 on Build GRCh38
rs539062649 Chr.22: 24541330-24541330 on Build GRCh38
rs758761377 Chr.22: 24541238-24541238 on Build GRCh38
rs1016164486 Chr.22: 24541368-24541368 on Build GRCh38
rs187228379 Chr.22: 24541400-24541400 on Build GRCh38
rs370564786 Chr.22: 24541422-24541422 on Build GRCh38
rs770720913 Chr.22: 24541202-24541214 on Build GRCh38
rs908219326 Chr.22: 24541470-24541470 on Build GRCh38
rs545961160 Chr.22: 24541487-24541487 on Build GRCh38
rs577142709 Chr.22: 24541546-24541546 on Build GRCh38
rs1045116440 Chr.22: 24541521-24541522 on Build GRCh38
rs1041003470 Chr.22: 24541629-24541633 on Build GRCh38
rs1028765414 Chr.22: 24541436-24541436 on Build GRCh38
rs901628778 Chr.22: 24541315-24541315 on Build GRCh38
rs892477279 Chr.22: 24541602-24541602 on Build GRCh38
rs1050591665 Chr.22: 24541591-24541591 on Build GRCh38
rs143077742 Chr.22: 24541584-24541584 on Build GRCh38
rs759207809 Chr.22: 24541633-24541634 on Build GRCh38
rs558066470 Chr.22: 24541394-24541394 on Build GRCh38
rs557139552 Chr.22: 24541442-24541442 on Build GRCh38
rs908148105 Chr.22: 24541227-24541227 on Build GRCh38
rs940850425 Chr.22: 24541259-24541259 on Build GRCh38
rs1008525619 Chr.22: 24541623-24541623 on Build GRCh38
rs1035902377 Chr.22: 24541484-24541484 on Build GRCh38
rs1056160301 Chr.22: 24541206-24541206 on Build GRCh38
rs2011947 Chr.22: 24541519-24541519 on Build GRCh38
rs139156002 Chr.22: 24541333-24541333 on Build GRCh38
rs997278976 Chr.22: 24541323-24541323 on Build GRCh38
rs1057009619 Chr.22: 24541260-24541260 on Build GRCh38
rs76845003 Chr.22: 24541630-24541630 on Build GRCh38
rs750234735 Chr.22: 24541279-24541279 on Build GRCh38
rs1030901225 Chr.22: 24541343-24541343 on Build GRCh38
rs903470965 Chr.22: 24541414-24541414 on Build GRCh38
rs984653403 Chr.22: 24541359-24541359 on Build GRCh38
rs761166394 Chr.22: 24541278-24541278 on Build GRCh38
rs1030748488 Chr.22: 24541309-24541309 on Build GRCh38
rs569324590 Chr.22: 24541292-24541292 on Build GRCh38
rs961895158 Chr.22: 24541456-24541456 on Build GRCh38
rs894482679 Chr.22: 24541248-24541248 on Build GRCh38
rs922743105 Chr.22: 24541568-24541568 on Build GRCh38
rs190907127 Chr.22: 24541420-24541420 on Build GRCh38
rs749198357 Chr.22: 24541486-24541486 on Build GRCh38
rs752973244 Chr.22: 24541228-24541228 on Build GRCh38
rs564411796 Chr.22: 24541493-24541493 on Build GRCh38
rs374598316 Chr.22: 24541591-24541592 on Build GRCh38
rs377185268 Chr.22: 24541628-24541629 on Build GRCh38
rs535165440 Chr.22: 24541441-24541441 on Build GRCh38
rs182666909 Chr.22: 24541560-24541560 on Build GRCh38
rs997650530 Chr.22: 24541430-24541430 on Build GRCh38
rs1005927994 Chr.22: 24541648-24541648 on Build GRCh38
rs529322933 Chr.22: 24541226-24541226 on Build GRCh38
rs961842936 Chr.22: 24541371-24541371 on Build GRCh38
rs5844595 Chr.22: 24541645-24541646 on Build GRCh38
rs888902197 Chr.22: 24541610-24541610 on Build GRCh38
rs903856459 Chr.22: 24541300-24541300 on Build GRCh38
rs764397325 Chr.22: 24541243-24541243 on Build GRCh38
rs200053659 Chr.22: 24541449-24541449 on Build GRCh38
rs928488614 Chr.22: 24541382-24541382 on Build GRCh38
rs947591813 Chr.22: 24541276-24541276 on Build GRCh38
rs971961170 Chr.22: 24541457-24541457 on Build GRCh38
rs938729511 Chr.22: 24541385-24541385 on Build GRCh38
rs953090965 Chr.22: 24541345-24541345 on Build GRCh38
rs991603029 Chr.22: 24541393-24541393 on Build GRCh38
rs551171479 Chr.22: 24541280-24541280 on Build GRCh38
rs924609635 Chr.22: 24541542-24541542 on Build GRCh38

View Full Product Details