Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.22: 41621184-41621693 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
XRCC6
Entrez Gene ID
2547
Gene Name
X-ray repair cross complementing 6
Gene Aliases
CTC75, CTCBF, G22P1, KU70, ML8, TLAA
UniGene
Hs.292493
Gene Chromosome Location
Chr.22: 41621163-41664048 on Build GRCh38
Transcripts
NM_001288976.1 NM_001288977.1 NM_001288978.1 NM_001469.4

SNP Detail

SNP ID Chromosome Location
rs922800546 Chr.22: 41621303-41621303 on Build GRCh38
rs979060907 Chr.22: 41621517-41621517 on Build GRCh38
rs779569430 Chr.22: 41621589-41621589 on Build GRCh38
rs1030177964 Chr.22: 41621646-41621646 on Build GRCh38
rs1006998869 Chr.22: 41621652-41621652 on Build GRCh38
rs772231151 Chr.22: 41621249-41621249 on Build GRCh38
rs11702892 Chr.22: 41621502-41621502 on Build GRCh38
rs36218276 Chr.22: 41621458-41621458 on Build GRCh38
rs999349162 Chr.22: 41621230-41621230 on Build GRCh38
rs72547477 Chr.22: 41621625-41621625 on Build GRCh38
rs750028122 Chr.22: 41621556-41621556 on Build GRCh38
rs1054521413 Chr.22: 41621643-41621643 on Build GRCh38
rs935111238 Chr.22: 41621341-41621341 on Build GRCh38
rs922797624 Chr.22: 41621445-41621445 on Build GRCh38
rs185292468 Chr.22: 41621509-41621509 on Build GRCh38
rs941964495 Chr.22: 41621579-41621579 on Build GRCh38
rs951437530 Chr.22: 41621366-41621366 on Build GRCh38
rs543994317 Chr.22: 41621328-41621328 on Build GRCh38
rs984153802 Chr.22: 41621371-41621371 on Build GRCh38
rs1003552510 Chr.22: 41621348-41621348 on Build GRCh38
rs376876886 Chr.22: 41621250-41621250 on Build GRCh38
rs759539018 Chr.22: 41621356-41621356 on Build GRCh38
rs753164206 Chr.22: 41621435-41621435 on Build GRCh38
rs972917607 Chr.22: 41621291-41621291 on Build GRCh38
rs529004245 Chr.22: 41621397-41621397 on Build GRCh38
rs865821422 Chr.22: 41621601-41621601 on Build GRCh38
rs76498295 Chr.22: 41621456-41621456 on Build GRCh38
rs961481581 Chr.22: 41621439-41621439 on Build GRCh38
rs923643480 Chr.22: 41621531-41621531 on Build GRCh38
rs532935670 Chr.22: 41621457-41621457 on Build GRCh38
rs752490256 Chr.22: 41621290-41621290 on Build GRCh38
rs193153213 Chr.22: 41621474-41621474 on Build GRCh38
rs997523488 Chr.22: 41621532-41621548 on Build GRCh38
rs1025656047 Chr.22: 41621544-41621544 on Build GRCh38
rs132770 Chr.22: 41621260-41621260 on Build GRCh38
rs1039922874 Chr.22: 41621298-41621298 on Build GRCh38
rs896397612 Chr.22: 41621246-41621246 on Build GRCh38
rs907143897 Chr.22: 41621669-41621669 on Build GRCh38
rs28384708 Chr.22: 41621649-41621649 on Build GRCh38
rs372109912 Chr.22: 41621406-41621406 on Build GRCh38
rs1018462196 Chr.22: 41621503-41621503 on Build GRCh38
rs118087986 Chr.22: 41621257-41621257 on Build GRCh38
rs910857586 Chr.22: 41621325-41621325 on Build GRCh38
rs1012137294 Chr.22: 41621394-41621394 on Build GRCh38
rs769645645 Chr.22: 41621663-41621663 on Build GRCh38
rs982076093 Chr.22: 41621258-41621258 on Build GRCh38
rs975574982 Chr.22: 41621438-41621438 on Build GRCh38
rs568980658 Chr.22: 41621498-41621498 on Build GRCh38
rs993311599 Chr.22: 41621540-41621540 on Build GRCh38
rs555174320 Chr.22: 41621465-41621465 on Build GRCh38
rs1005945486 Chr.22: 41621560-41621560 on Build GRCh38
rs28384707 Chr.22: 41621276-41621276 on Build GRCh38
rs7284074 Chr.22: 41621662-41621662 on Build GRCh38
rs975627597 Chr.22: 41621578-41621578 on Build GRCh38
rs757600573 Chr.22: 41621549-41621549 on Build GRCh38
rs1016973492 Chr.22: 41621561-41621561 on Build GRCh38
rs746927221 Chr.22: 41621275-41621275 on Build GRCh38
rs531853481 Chr.22: 41621302-41621302 on Build GRCh38
rs36217922 Chr.22: 41621258-41621258 on Build GRCh38
rs764550273 Chr.22: 41621508-41621508 on Build GRCh38
rs1044944039 Chr.22: 41621569-41621569 on Build GRCh38
rs386821478 Chr.22: 41621258-41621260 on Build GRCh38
rs935653776 Chr.22: 41621484-41621484 on Build GRCh38
rs1045634521 Chr.22: 41621525-41621525 on Build GRCh38
rs547477779 Chr.22: 41621415-41621415 on Build GRCh38
rs1029794186 Chr.22: 41621487-41621487 on Build GRCh38
rs961788070 Chr.22: 41621279-41621279 on Build GRCh38
rs902817181 Chr.22: 41621316-41621316 on Build GRCh38
rs947889320 Chr.22: 41621557-41621557 on Build GRCh38
rs946432727 Chr.22: 41621216-41621216 on Build GRCh38
rs910110464 Chr.22: 41621386-41621386 on Build GRCh38
rs923701791 Chr.22: 41621337-41621337 on Build GRCh38
rs553830742 Chr.22: 41621658-41621658 on Build GRCh38
rs1050385438 Chr.22: 41621557-41621573 on Build GRCh38
rs778554475 Chr.22: 41621552-41621552 on Build GRCh38
rs28384706 Chr.22: 41621243-41621243 on Build GRCh38
rs776617584 Chr.22: 41621304-41621304 on Build GRCh38
rs946401689 Chr.22: 41621326-41621326 on Build GRCh38
rs555548472 Chr.22: 41621615-41621615 on Build GRCh38
rs132770 Chr.22: 41621260-41621260 on Build GRCh38
rs568516634 Chr.22: 41621393-41621393 on Build GRCh38
rs11557350 Chr.22: 41621320-41621320 on Build GRCh38
rs77728367 Chr.22: 41621207-41621207 on Build GRCh38
rs112105277 Chr.22: 41621333-41621333 on Build GRCh38
rs571540057 Chr.22: 41621226-41621226 on Build GRCh38
rs573923013 Chr.22: 41621240-41621240 on Build GRCh38
rs377088635 Chr.22: 41621315-41621315 on Build GRCh38
rs1005538785 Chr.22: 41621425-41621425 on Build GRCh38
rs536903335 Chr.22: 41621573-41621573 on Build GRCh38
rs964635154 Chr.22: 41621564-41621564 on Build GRCh38
rs930002840 Chr.22: 41621259-41621259 on Build GRCh38
rs767593415 Chr.22: 41621409-41621409 on Build GRCh38
rs17002533 Chr.22: 41621350-41621350 on Build GRCh38
rs1043467338 Chr.22: 41621224-41621224 on Build GRCh38
rs36218275 Chr.22: 41621313-41621313 on Build GRCh38

View Full Product Details