Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 10325990-10326504 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ATP2B2
Entrez Gene ID
491
Gene Name
ATPase plasma membrane Ca2+ transporting 2
Gene Aliases
PMCA2, PMCA2a, PMCA2i
UniGene
Hs.268942
Gene Chromosome Location
Chr.3: 10324023-10707962 on Build GRCh38
Transcripts
NM_001001331.2 NM_001683.3 XM_005265179.4 XM_006713175.3 XM_011533752.2 XM_017006481.1 XM_017006482.1 XM_017006483.1 XM_017006484.1 XM_017006485.1 XM_017006486.1 XM_017006487.1 XM_017006489.1 XM_017006491.1 XM_017006492.1

SNP Detail

SNP ID Chromosome Location
rs758560258 Chr.3: 10326346-10326346 on Build GRCh38
rs184630068 Chr.3: 10326259-10326259 on Build GRCh38
rs14154 Chr.3: 10326429-10326429 on Build GRCh38
rs879040734 Chr.3: 10326040-10326040 on Build GRCh38
rs925745830 Chr.3: 10326356-10326356 on Build GRCh38
rs117060922 Chr.3: 10326414-10326414 on Build GRCh38
rs763480317 Chr.3: 10326260-10326261 on Build GRCh38
rs191433362 Chr.3: 10326180-10326180 on Build GRCh38
rs760410282 Chr.3: 10326196-10326196 on Build GRCh38
rs887017507 Chr.3: 10326478-10326478 on Build GRCh38
rs1055387577 Chr.3: 10326181-10326181 on Build GRCh38
rs752770830 Chr.3: 10326337-10326337 on Build GRCh38
rs1042748900 Chr.3: 10326079-10326079 on Build GRCh38
rs780288418 Chr.3: 10326259-10326260 on Build GRCh38
rs895765085 Chr.3: 10326206-10326206 on Build GRCh38
rs188313558 Chr.3: 10326306-10326306 on Build GRCh38
rs891977862 Chr.3: 10326258-10326258 on Build GRCh38
rs914045854 Chr.3: 10326038-10326038 on Build GRCh38
rs58208242 Chr.3: 10326438-10326438 on Build GRCh38
rs3194714 Chr.3: 10326207-10326207 on Build GRCh38
rs113724179 Chr.3: 10326271-10326274 on Build GRCh38
rs773160018 Chr.3: 10326081-10326081 on Build GRCh38
rs369886126 Chr.3: 10326204-10326204 on Build GRCh38
rs925002571 Chr.3: 10326477-10326477 on Build GRCh38
rs35126324 Chr.3: 10326098-10326099 on Build GRCh38
rs3194712 Chr.3: 10326291-10326291 on Build GRCh38
rs111994765 Chr.3: 10326098-10326098 on Build GRCh38
rs150569707 Chr.3: 10326193-10326193 on Build GRCh38
rs371215484 Chr.3: 10326258-10326259 on Build GRCh38
rs1029318971 Chr.3: 10326464-10326464 on Build GRCh38
rs59344515 Chr.3: 10326245-10326248 on Build GRCh38
rs988744173 Chr.3: 10326021-10326021 on Build GRCh38
rs192867489 Chr.3: 10326370-10326370 on Build GRCh38
rs41147 Chr.3: 10326418-10326418 on Build GRCh38
rs1001932670 Chr.3: 10326369-10326369 on Build GRCh38
rs925631974 Chr.3: 10326156-10326156 on Build GRCh38
rs975965936 Chr.3: 10326456-10326456 on Build GRCh38
rs139286238 Chr.3: 10326243-10326246 on Build GRCh38
rs965937011 Chr.3: 10326055-10326055 on Build GRCh38
rs556098142 Chr.3: 10326469-10326469 on Build GRCh38
rs945804492 Chr.3: 10326048-10326048 on Build GRCh38
rs937045476 Chr.3: 10326168-10326168 on Build GRCh38
rs556983530 Chr.3: 10326259-10326262 on Build GRCh38
rs752135322 Chr.3: 10326412-10326412 on Build GRCh38
rs577533345 Chr.3: 10326020-10326020 on Build GRCh38
rs968212916 Chr.3: 10326333-10326337 on Build GRCh38
rs113606917 Chr.3: 10326100-10326100 on Build GRCh38
rs201372088 Chr.3: 10326246-10326249 on Build GRCh38
rs1006058090 Chr.3: 10326235-10326236 on Build GRCh38
rs925022625 Chr.3: 10326229-10326229 on Build GRCh38
rs887839928 Chr.3: 10326407-10326407 on Build GRCh38
rs921578395 Chr.3: 10326158-10326158 on Build GRCh38
rs557855071 Chr.3: 10326262-10326263 on Build GRCh38
rs113958840 Chr.3: 10326097-10326097 on Build GRCh38
rs931649814 Chr.3: 10326177-10326177 on Build GRCh38
rs537736232 Chr.3: 10326467-10326467 on Build GRCh38
rs397830756 Chr.3: 10326099-10326100 on Build GRCh38
rs112255964 Chr.3: 10326245-10326245 on Build GRCh38
rs186955709 Chr.3: 10326152-10326152 on Build GRCh38
rs935053345 Chr.3: 10326479-10326479 on Build GRCh38
rs572088509 Chr.3: 10326460-10326460 on Build GRCh38
rs752915391 Chr.3: 10326417-10326417 on Build GRCh38
rs935120197 Chr.3: 10326231-10326231 on Build GRCh38

View Full Product Details