Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 50613650-50614185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MAPKAPK3
Entrez Gene ID
7867
Gene Name
mitogen-activated protein kinase-activated protein kinase 3
Gene Aliases
3PK, MAPKAP-K3, MAPKAP3, MAPKAPK-3, MK-3
UniGene
Hs.234521
Gene Chromosome Location
Chr.3: 50611862-50649297 on Build GRCh38
Transcripts
NM_001243926.1

SNP Detail

SNP ID Chromosome Location
rs140793721 Chr.3: 50614024-50614024 on Build GRCh38
rs550878308 Chr.3: 50613884-50613884 on Build GRCh38
rs935911051 Chr.3: 50613825-50613825 on Build GRCh38
rs759998067 Chr.3: 50613828-50613828 on Build GRCh38
rs1025758865 Chr.3: 50614014-50614014 on Build GRCh38
rs993206470 Chr.3: 50613975-50613975 on Build GRCh38
rs777267881 Chr.3: 50613771-50613771 on Build GRCh38
rs1033993471 Chr.3: 50614086-50614086 on Build GRCh38
rs898839692 Chr.3: 50613967-50613967 on Build GRCh38
rs150954973 Chr.3: 50613969-50613969 on Build GRCh38
rs753165048 Chr.3: 50614110-50614110 on Build GRCh38
rs541670482 Chr.3: 50613996-50613996 on Build GRCh38
rs925611836 Chr.3: 50614146-50614146 on Build GRCh38
rs75701766 Chr.3: 50613965-50613965 on Build GRCh38
rs549731861 Chr.3: 50614006-50614006 on Build GRCh38
rs947447918 Chr.3: 50613819-50613819 on Build GRCh38
rs1038713480 Chr.3: 50613781-50613781 on Build GRCh38
rs369276260 Chr.3: 50613914-50613914 on Build GRCh38
rs981968650 Chr.3: 50614084-50614084 on Build GRCh38
rs933530430 Chr.3: 50613872-50613872 on Build GRCh38
rs1052440123 Chr.3: 50613943-50613943 on Build GRCh38
rs955565890 Chr.3: 50614053-50614053 on Build GRCh38
rs568003581 Chr.3: 50613692-50613692 on Build GRCh38
rs569006069 Chr.3: 50613944-50613944 on Build GRCh38
rs959292552 Chr.3: 50614108-50614108 on Build GRCh38
rs989818969 Chr.3: 50614125-50614125 on Build GRCh38
rs914577007 Chr.3: 50613802-50613802 on Build GRCh38
rs987217700 Chr.3: 50614079-50614079 on Build GRCh38
rs150449425 Chr.3: 50614033-50614033 on Build GRCh38
rs743753 Chr.3: 50613964-50613964 on Build GRCh38
rs541893477 Chr.3: 50614137-50614137 on Build GRCh38
rs535128073 Chr.3: 50613694-50613694 on Build GRCh38
rs991526667 Chr.3: 50613790-50613790 on Build GRCh38
rs377066480 Chr.3: 50613993-50613993 on Build GRCh38
rs145075748 Chr.3: 50613853-50613853 on Build GRCh38

View Full Product Details