Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 169773356-169773853 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
MYNN
Entrez Gene ID
55892
Gene Name
myoneurin
Gene Aliases
OSZF, SBBIZ1, ZBTB31, ZNF902
UniGene
Hs.507025
Gene Chromosome Location
Chr.3: 169772247-169789716 on Build GRCh38
Transcripts
NM_001185118.1 NM_018657.4 NR_033702.1 NR_033703.1 XM_005247621.4 XM_005247622.3 XM_005247624.3 XM_011512987.1 XM_017006864.1 XM_017006865.1 XM_017006866.1 XM_017006867.1 XM_017006868.1

SNP Detail

SNP ID Chromosome Location
rs924184821 Chr.3: 169773784-169773784 on Build GRCh38
rs982413114 Chr.3: 169773813-169773813 on Build GRCh38
rs879706842 Chr.3: 169773740-169773740 on Build GRCh38
rs1027316449 Chr.3: 169773757-169773757 on Build GRCh38
rs998804154 Chr.3: 169773621-169773621 on Build GRCh38
rs968295591 Chr.3: 169773779-169773779 on Build GRCh38
rs761861680 Chr.3: 169773749-169773749 on Build GRCh38
rs1009413810 Chr.3: 169773649-169773649 on Build GRCh38
rs140519498 Chr.3: 169773516-169773516 on Build GRCh38
rs886882639 Chr.3: 169773400-169773400 on Build GRCh38
rs954825695 Chr.3: 169773437-169773438 on Build GRCh38
rs908211680 Chr.3: 169773817-169773817 on Build GRCh38
rs567088620 Chr.3: 169773783-169773783 on Build GRCh38
rs1018440129 Chr.3: 169773375-169773375 on Build GRCh38
rs904508044 Chr.3: 169773425-169773425 on Build GRCh38
rs1021263725 Chr.3: 169773750-169773750 on Build GRCh38
rs908285389 Chr.3: 169773515-169773515 on Build GRCh38
rs975296984 Chr.3: 169773743-169773743 on Build GRCh38
rs948941335 Chr.3: 169773542-169773542 on Build GRCh38
rs904821628 Chr.3: 169773593-169773593 on Build GRCh38
rs964107826 Chr.3: 169773728-169773728 on Build GRCh38
rs1056591303 Chr.3: 169773459-169773459 on Build GRCh38
rs532038399 Chr.3: 169773446-169773446 on Build GRCh38
rs939556952 Chr.3: 169773785-169773785 on Build GRCh38
rs1019415050 Chr.3: 169773693-169773693 on Build GRCh38
rs1057263706 Chr.3: 169773522-169773522 on Build GRCh38
rs946931398 Chr.3: 169773405-169773405 on Build GRCh38
rs552625234 Chr.3: 169773815-169773815 on Build GRCh38
rs919883634 Chr.3: 169773471-169773471 on Build GRCh38
rs1057492539 Chr.3: 169773409-169773409 on Build GRCh38
rs986562535 Chr.3: 169773437-169773437 on Build GRCh38
rs527873862 Chr.3: 169773797-169773797 on Build GRCh38
rs894695183 Chr.3: 169773419-169773419 on Build GRCh38
rs1038303546 Chr.3: 169773517-169773517 on Build GRCh38
rs529819424 Chr.3: 169773603-169773603 on Build GRCh38
rs959747383 Chr.3: 169773830-169773830 on Build GRCh38
rs1025508739 Chr.3: 169773588-169773588 on Build GRCh38
rs990211518 Chr.3: 169773383-169773383 on Build GRCh38
rs570474446 Chr.3: 169773537-169773537 on Build GRCh38
rs895201473 Chr.3: 169773461-169773461 on Build GRCh38
rs1021595943 Chr.3: 169773424-169773424 on Build GRCh38
rs1013602520 Chr.3: 169773423-169773423 on Build GRCh38
rs916681994 Chr.3: 169773810-169773810 on Build GRCh38
rs994005482 Chr.3: 169773579-169773579 on Build GRCh38
rs1008460437 Chr.3: 169773662-169773662 on Build GRCh38
rs930948343 Chr.3: 169773479-169773479 on Build GRCh38
rs1031639289 Chr.3: 169773629-169773629 on Build GRCh38
rs978260913 Chr.3: 169773412-169773412 on Build GRCh38
rs114340092 Chr.3: 169773646-169773646 on Build GRCh38
rs963770702 Chr.3: 169773380-169773380 on Build GRCh38
rs915511279 Chr.3: 169773396-169773396 on Build GRCh38
rs763458750 Chr.3: 169773422-169773422 on Build GRCh38
rs982485313 Chr.3: 169773490-169773490 on Build GRCh38
rs562304501 Chr.3: 169773572-169773572 on Build GRCh38
rs1018053651 Chr.3: 169773716-169773716 on Build GRCh38
rs998833857 Chr.3: 169773435-169773435 on Build GRCh38
rs939505313 Chr.3: 169773447-169773447 on Build GRCh38
rs769114516 Chr.3: 169773483-169773483 on Build GRCh38
rs938831952 Chr.3: 169773402-169773402 on Build GRCh38
rs931021226 Chr.3: 169773388-169773388 on Build GRCh38
rs1031714212 Chr.3: 169773436-169773436 on Build GRCh38

View Full Product Details