Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 129109375-129109919 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C4orf33
Entrez Gene ID
132321
Gene Name
chromosome 4 open reading frame 33
Gene Aliases
-
UniGene
Hs.567679
Gene Chromosome Location
Chr.4: 129093674-129112688 on Build GRCh38
Transcripts
NM_001099783.1 NM_173487.2 XM_005262734.4 XM_017007722.1 XM_017007723.1 XM_017007724.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4122250 c.494+2T>G p.? Chr.4: 129109674-129109674 on Build GRCh38
6202922 c.437A>G p.Y146C Chr.4: 129109615-129109615 on Build GRCh38
6099352 c.367A>T p.S123C Chr.4: 129109545-129109545 on Build GRCh38
4122249 c.474G>C p.Q158H Chr.4: 129109652-129109652 on Build GRCh38
1179821 c.450C>T p.Y150Y Chr.4: 129109628-129109628 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs2271571 Chr.4: 129109541-129109541 on Build GRCh38
rs376154093 Chr.4: 129109580-129109580 on Build GRCh38
rs1042280544 Chr.4: 129109696-129109696 on Build GRCh38
rs372722498 Chr.4: 129109643-129109643 on Build GRCh38
rs528534972 Chr.4: 129109782-129109782 on Build GRCh38
rs3105367 Chr.4: 129109666-129109666 on Build GRCh38
rs1012160025 Chr.4: 129109768-129109768 on Build GRCh38
rs759642263 Chr.4: 129109433-129109433 on Build GRCh38
rs337276 Chr.4: 129109789-129109789 on Build GRCh38
rs758915998 Chr.4: 129109720-129109720 on Build GRCh38
rs976067808 Chr.4: 129109863-129109863 on Build GRCh38
rs10009430 Chr.4: 129109608-129109608 on Build GRCh38
rs760945668 Chr.4: 129109483-129109483 on Build GRCh38
rs776865920 Chr.4: 129109611-129109611 on Build GRCh38
rs369880865 Chr.4: 129109424-129109424 on Build GRCh38
rs999151818 Chr.4: 129109437-129109437 on Build GRCh38
rs1033606952 Chr.4: 129109574-129109574 on Build GRCh38
rs560230863 Chr.4: 129109812-129109812 on Build GRCh38
rs749633073 Chr.4: 129109584-129109584 on Build GRCh38
rs574654514 Chr.4: 129109725-129109725 on Build GRCh38
rs771536546 Chr.4: 129109408-129109408 on Build GRCh38
rs759508018 Chr.4: 129109636-129109636 on Build GRCh38
rs372176189 Chr.4: 129109710-129109710 on Build GRCh38
rs774218552 Chr.4: 129109694-129109694 on Build GRCh38
rs182346325 Chr.4: 129109609-129109609 on Build GRCh38
rs974557612 Chr.4: 129109577-129109577 on Build GRCh38
rs763791839 Chr.4: 129109656-129109656 on Build GRCh38
rs767408001 Chr.4: 129109640-129109640 on Build GRCh38
rs747070649 Chr.4: 129109726-129109726 on Build GRCh38
rs762684903 Chr.4: 129109612-129109612 on Build GRCh38
rs961345279 Chr.4: 129109642-129109642 on Build GRCh38
rs183442181 Chr.4: 129109860-129109860 on Build GRCh38
rs772206461 Chr.4: 129109448-129109448 on Build GRCh38
rs749722760 Chr.4: 129109403-129109403 on Build GRCh38
rs879084754 Chr.4: 129109514-129109514 on Build GRCh38
rs2271570 Chr.4: 129109489-129109489 on Build GRCh38
rs906383356 Chr.4: 129109532-129109532 on Build GRCh38
rs986730913 Chr.4: 129109602-129109602 on Build GRCh38
rs767054190 Chr.4: 129109555-129109555 on Build GRCh38
rs998103129 Chr.4: 129109891-129109891 on Build GRCh38
rs191190723 Chr.4: 129109832-129109832 on Build GRCh38
rs762774966 Chr.4: 129109490-129109490 on Build GRCh38
rs369274698 Chr.4: 129109585-129109585 on Build GRCh38
rs1033969001 Chr.4: 129109820-129109820 on Build GRCh38
rs1025768015 Chr.4: 129109594-129109594 on Build GRCh38
rs576609928 Chr.4: 129109713-129109714 on Build GRCh38
rs1056117271 Chr.4: 129109752-129109752 on Build GRCh38
rs337277 Chr.4: 129109497-129109497 on Build GRCh38
rs755773616 Chr.4: 129109564-129109564 on Build GRCh38
rs764118985 Chr.4: 129109756-129109756 on Build GRCh38
rs544444481 Chr.4: 129109633-129109633 on Build GRCh38
rs754864944 Chr.4: 129109605-129109605 on Build GRCh38
rs185150316 Chr.4: 129109769-129109769 on Build GRCh38
rs147330290 Chr.4: 129109563-129109563 on Build GRCh38
rs912576051 Chr.4: 129109604-129109604 on Build GRCh38
rs375827246 Chr.4: 129109399-129109399 on Build GRCh38
rs532645909 Chr.4: 129109842-129109842 on Build GRCh38
rs757629574 Chr.4: 129109695-129109695 on Build GRCh38
rs114250643 Chr.4: 129109628-129109628 on Build GRCh38
rs779415757 Chr.4: 129109709-129109709 on Build GRCh38
rs936279609 Chr.4: 129109728-129109728 on Build GRCh38
rs373056399 Chr.4: 129109430-129109430 on Build GRCh38
rs751518318 Chr.4: 129109508-129109508 on Build GRCh38
rs572798457 Chr.4: 129109598-129109598 on Build GRCh38
rs554326745 Chr.4: 129109591-129109591 on Build GRCh38
rs562691711 Chr.4: 129109671-129109671 on Build GRCh38
rs769403563 Chr.4: 129109400-129109400 on Build GRCh38
rs1001951328 Chr.4: 129109802-129109802 on Build GRCh38
rs764279956 Chr.4: 129109487-129109487 on Build GRCh38
rs781089927 Chr.4: 129109677-129109677 on Build GRCh38
rs1023868736 Chr.4: 129109519-129109519 on Build GRCh38
rs778191552 Chr.4: 129109401-129109401 on Build GRCh38
rs961377549 Chr.4: 129109884-129109884 on Build GRCh38
rs772521420 Chr.4: 129109600-129109600 on Build GRCh38
rs150941780 Chr.4: 129109646-129109646 on Build GRCh38
rs747119304 Chr.4: 129109459-129109461 on Build GRCh38
rs561181487 Chr.4: 129109770-129109770 on Build GRCh38
rs775509416 Chr.4: 129109460-129109460 on Build GRCh38
rs1024699653 Chr.4: 129109530-129109530 on Build GRCh38

View Full Product Details